Invitrogen™

SureCast™ Acrylamide Solution (40%)

Catalog number: HC2040
Have Questions?
Invitrogen™

SureCast™ Acrylamide Solution (40%)

Catalog number: HC2040
Have Questions?
Catalog Number
HC2040
Unit Size
450 mL
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
HC2040450 mL
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
SureCast Acrylamide Solution (40%) can be used to prepare single-percentage and gradient gels using the SureCast Handcast System or in other hand-casting systems. Acrylamide is a major ingredient in polyacrylamide gels used for separating proteins during polyacrylamide gel electrophoresis (PAGE).

Features of the SureCast Acrylamide Solution include:
• Bis-acrylamide ratio of 29:1
• Room temperature storage
• Long shelf life
• High purity
• Safer alternative to powdered acrylamide
• Concentrated to enable the hand-casting of a broad range of gel percentages

See all gel casting reagents ›
Learn more about the SureCast Handcast system ›

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Product Line
SureCast
Type
40% Acrylamide Solution
Shipping Condition
Room Temperature

Contents & Storage

Store at room temperature.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text