Invitrogen™

SureCast™ Stacking Buffer

Catalog number: HC2112
Have Questions?
Invitrogen™

SureCast™ Stacking Buffer

Catalog number: HC2112
Have Questions?
Catalog Number
HC2112
Unit Size
2 x 500 mL
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
5 x 500 mL
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
HC21122 x 500 mL
-
HC21155 x 500 mL
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
SureCast Stacking Buffer packs are pouches of dry blend powder, each sufficient to make 500 mL of stacking gel buffer (0.5 M Tris-HCl buffer, pH 6.8) for hand-casting polyacrylamide gels. They are easy to use—simply empty the contents of one pouch into a beaker, add ultrapure water, and stir to dissolve.

See all gel casting reagents ›
Learn more about the SureCast Handcast system ›

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Product Type
Stacking Buffer (1 L)
Product Line
SureCast™
Quantity
2 x 500 mL
Shipping Condition
Room Temperature

Contents & Storage

2 x 500 mL (1 L total) SureCast Stacking Buffer packs.
  • Store at room temperature.
  • Figures

    Documents & Downloads

    Certificates

      Frequently asked questions (FAQs)

      Citations & References

      Search citations by name, author, journal title or abstract text