Activated Human Myofibroblastic Hepatic Stellate Cells (MF-HSCs)
Activated Human Myofibroblastic Hepatic Stellate Cells (MF-HSCs)
Gibco™

Activated Human Myofibroblastic Hepatic Stellate Cells (MF-HSCs)

Gibco Activated Human Myofibroblastic Hepatic Stellate Cells (MF-HSCs) are produced from isolated pure quiescent human stellate cells (q- HSCs). TheRead more
Have Questions?
Catalog NumberQuantity
HMFHSC1 Vial
Catalog number HMFHSC
Price (USD)
2,400.00
Each
-
Add to cart
Quantity:
1 Vial
Price (USD)
2,400.00
Each
Add to cart
Ask our AI about this Product
Gibco Activated Human Myofibroblastic Hepatic Stellate Cells (MF-HSCs) are produced from isolated pure quiescent human stellate cells (q- HSCs). The cells are obtained from a single donor and provided in a cryopreserved format identical to single-donor plateable human hepatocytes.

Each vial contains a minimum of one million viable cells at passage 5, sufficient for at least four 96-well plates in 2D culture.

Simplified protocol
Activated MF-HSCs are suitable for building hepatic co-culture with primary hepatocytes to study in vitro disease models such as hepatic fibrosis. Cryopreserved cells can be thawed and plated at your convenience in as little as 30 minutes. Try it on your own, or request a demo with an experienced scientist.

Stringent quality control
Each lot is tested for:
• Viability >75%
• Attachment efficiency
• Proper morphology
• Purity based on α-smooth muscle actin (α-SMA)
• Activation in response to Transforming Growth Factor-β1 (TGF-β1)
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Donor SourceSingle Donor
Physical FormCryopreserved
Quantity1 Vial
Host SpeciesHuman
Product TypeMyofibroblastic Hepatic Stellate Cells (MF-HSCs)
Shipping ConditionDry shipper
Unit SizeEach
Contents & Storage
>1 million viable cells/vial
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link