Gibco™

Human Microsomes, 50 Donors

Catalog number: HMMCPL
Have Questions?
Gibco™

Human Microsomes, 50 Donors

Catalog number: HMMCPL
Have Questions?
Catalog Number
HMMCPL
Unit Size
0.5 mL
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
HMMCPL0.5 mL
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Human liver microsomes pooled from 50 different individual donors. All donors are equally represented for a truer population sample. Fully characterized for major cytochrome P450 activities and select Phase II enzymes.

Total protein concentration: 20 mg/ml

We provide liver subcellular fractions from a variety of tox species, including human, nonhuman primates (Cynomolgus Monkey and Rhesus Monkey), dog (Beagle), rat (Sprague-Dawley), mouse (CD-1 and Balb-c), and trout (Oncorhynchus mykiss). Custom species are available upon request.

For research use only. Not intended for human or animal therapeutic or diagnostic use.

Related Link

Learn more about our microsomes and subcellular fractions
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Donor Source
Pooled
Gender
Unisex
Physical Form
Frozen
Tissue
Liver
Quantity
0.5 mL
Purity
Research Grade
Host Species
Human
Concentration
20 mg/mL
Product Type
Human Microsome
Life Stage
Adult
Format
Tube
Sub Cellular Localization
Microsomes
Shipping Condition
Dry Ice
Product Line
Gibco™
Unit Size
0.5 mL

Contents & Storage

Store in ultracold freezer (-60 to -85°C) and protect from light.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text