Gibco™

HepaRG™ Cells, Cryopreserved

Catalog number: HPRGC10
Have Questions?
Gibco™

HepaRG™ Cells, Cryopreserved

Catalog number: HPRGC10
Have Questions?
Catalog Number
HPRGC10
Unit Size
10 x 106 cells
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
HPRGC1010 x 106 cells
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
The HepaRG™ cell line is an immortalized hepatic cell line that retains many characteristics of primary human hepatocytes. HepaRG™ cells are terminally differentiated and provided in a convenient cryopreserved format. For scientists who need reproducible metabolism data, HepaRG™ cells are an in vitro tool that provides reproducible results in a metabolically complete and scalable system.
• Obtain biologically relevant results from a metabolically complete system
• Assess the drug-drug interaction potential of your compound
• Experience reproducible results from a single population of cells

Biologically Relevant
HepaRG™ cells exhibit many characteristics of primary human hepatocytes including morphology, expression of key metabolic enzymes, expression of nuclear receptors, and drug transporters. Unlike HepG2 and Fa2N-4 cells, HepaRG™ cells have high P450 activity and complete expression of all nuclear receptors.

Predict Metabolism-based Drug-drug Interaction
HepaRG™ cells respond to prototypical P450 inducers and inhibitors to the same extent as primary hepatocytes, allowing HepaRG™ cells to be used to evaluate potential drug-drug interactions.

Infinitely Reproducible
HepaRG™ cells exhibit many characteristics of primary human hepatocytes and are essentially a single donor. These features allow users to obtain physiological relevant results for metabolism-based drug-drug interactions without the concern of donor variability and limited lot sizes that come with relying on donor tissue. (Note: The cells we provide are terminally differentiated).

HepaRG™ is a trademark of BioPredic International.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Age
Adult
Cell Line
HepaRG
Cell Type
Hepatocytes
Form
Cryopreserved
Gender
Female
Species
Human
Donor Source
Single Donor
No. of Cells
10 x 10^6
Product Type
Hepatocytes
Quantity
1 vial(s)
Shipping Condition
Dry Ice

Contents & Storage

1 vial, store in cryostorage dewar or -135°C freezer

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text