Search Thermo Fisher Scientific
The Thermo Scientific Helios Hydra PFIB-SEMs are versatile multi-application tools with four different ion species (argon, nitrogen, oxygen, and xenon), allowing you to choose the ions that provide the best results. Great results start with comprehensive sample preparation, whether you are performing S/TEM lamella imaging, 3D characterization, or large area physical failure analysis.
Now with enhanced, targeted applications capability, the Helios 5+ Hydra is here. Learn more about what this innovative system can do for your failure analysis and materials characterization challenges.
For Research Use Only. Not for use in diagnostic procedures.