Invitrogen™

Fixative-Free Lysing Solution, High-Yield Lyse

Catalog number: HYL250
Have Questions?
Invitrogen™

Fixative-Free Lysing Solution, High-Yield Lyse

Catalog number: HYL250
Have Questions?
Catalog Number
HYL250
Unit Size
500 mL
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
HYL250500 mL
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
High-Yield Lyse is a premixed, fixative-free erythrocyte lysing solution that can be used to eliminate red cells from whole blood for flow cytometric analysis with minimal loss of rare blood cell populations. Using this reagent, lysis of erythrocytes is performed immediately following staining of the blood samples with Invitrogen's fluorochrome-conjugated monoclonal antibodies.

View a selection guide for all cell lysis and preservation solutions and products related to sample fixation and permeablization, blocking reagents, and information about the use of Dynabeads™ for cell isolation.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Sample Type
Blood
Product Type
Lysing Solution
Quantity
500 mL
Reagent Type
Whole Blood Lysis Buffer
Shipping Condition
Room Temperature

Contents & Storage

Store at room temperature.

Shelf life: 24 months from date of manufacture

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text