Search Thermo Fisher Scientific
Applications | Building Materials, Coating and Converting |
Description | Extrusion Coating Application Package - Single Frame |
Unit Size | Each |
Applications | Coating and Converting |
Description | Roll Coating Application Package - Single Frame |
Unit Size | Each |
Catalog Number | Specifications | Unit Size | Description | Price (USD) | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
I-APP00-03 | Each | Extrusion Coating Application Package - Single Frame | Request A Quote | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
I-APP00-02 | Each | Roll Coating Application Package - Single Frame | Request A Quote | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|
The IPlus! Measurement and Control System for Coating and Converting combines advanced sensor technology and powerful software in an intuitive, easy-to-use system to provide high performance and a low cost of ownership. The system incorporates advanced application packages for converted product manufacturing to increase the quality, consistency and performance characteristics of coated and converted products.
The suite of Thermo Scientific advanced machine-direction and cross-direction controls improve both quality and productivity, delivering consistent results and an impressive return on investment. The Machine Direction (MD) thickness control option is used to automatically maintain the extruded or coated product to thickness specifications, while the Automatic Profile Control (APC) option controls auto dies to achieve cross-direction uniformity of extrusion coated products. Two-zone CD (Cross Direction) control improves the cross width profile uniformity of blade or roll coated products.
Features:
Operator Interface:
Advanced Wonderware™ user interface with Windows operating system offers easier data management and archiving. Desktop interface and/or console configurations are available.