Image-iT™ Green Hypoxia Reagent
Image-iT™ Green Hypoxia Reagent
Invitrogen™

Image-iT™ Green Hypoxia Reagent

Image-iT Green Hypoxia Reagent is a novel, fixable fluorogenic compound for measuring hypoxia in live cells. It is non-fluorescent whenRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
I148341 Vial
I148335 Vials
Catalog number I14834
Price (USD)
173.65
Online Exclusive
185.00
Save 11.35 (6%)
Each
Add to cart
Quantity:
1 Vial
Price (USD)
173.65
Online Exclusive
185.00
Save 11.35 (6%)
Each
Add to cart

Image-iT Green Hypoxia Reagent is a novel, fixable fluorogenic compound for measuring hypoxia in live cells. It is non-fluorescent when live cells are in an environment with normal oxygen concentrations and becomes fluorescent when oxygen levels are decreased. Image-iT Green Hypoxia Reagent sustains its fluorescence when cells/tissue return to normal oxygen levels, allowing the cells/tissue to be fixed with minimal loss of fluorescent signal.

Features of Image-iT Green Hypoxia Reagent include:

  • Indicates hypoxia in live cells by fluorescing in low oxygen environments
  • Fixable—sustains fluorescent signal when cells/tissue return to normal oxygen levels
  • Easy to use—just add to cell culture media and image

Image-iT Green Hypoxia Reagent is a fluorogenic compound that is live cell permeable and becomes fluorescent in environments with low oxygen concentrations. These properties makes it a highly useful tool for detecting cells and tissue under hypoxic conditions. Image-iT Green Hypoxia Reagent is a very sensitive oxygen detector. Unlike pimonidazole adducts that respond only to very low oxygen levels, Image-iT Green Hypoxia Reagent begins to fluoresce when atmospheric oxygen levels are less than 5%. It responds quickly to such environments, and the fluorescence is sustained after the oxygen levels return to normal. These properties make Image-iT Green Hypoxia Reagent an ideal tool for detecting hypoxic conditions around tumors, 3D cultures, spheroids, neurons, etc.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Quantity1 Vial
Reagent TypeHypoxia Reagent
Shipping ConditionApproved for shipment on Wet or Dry Ice
Product LineImage-iT
Product TypeReagent
Unit SizeEach
Contents & Storage
Store at -5 to -30°C. Protect from light.
  • Stable for at least 6 months after receiving.
  • Frequently asked questions (FAQs)

    Can Image-it Green Hypoxia Reagent be used in cells expressing fluorescent proteins?

    Yes, as long as the fluorescent protein does not emit in the green range (˜510–535 nm). For cells that do not express a fluorescent protein, we recommend viewing unstained cells under the FITC channel to examine autofluorescence. For cells expressing a fluorescent protein, we recommend analyzing unstained cell samples to determine the extent the fluorescent protein emission may overlap in the FITC channel.

    Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

    Our Image-iT Green Hypoxia Reagent is already highly fluorescent before using the product. What could have caused this?

    This could be due to storage of the product under anaerobic or low oxygen conditions. The Image-iT Green Hypoxia Reagent increases in fluorescence upon exposure to low oxygen environments, and this change is not reversible. For some ROS indicators, we recommend storing the reagent under dry nitrogen or argon to prevent oxidation during storage. However, this is not appropriate for the Image-iT Green Hypoxia Reagent.

    Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

    What is different between the use of the Hypoxia Green Reagent for Flow Cytometry (Cat No. H20035) versus imaging using the Image-iT Green Hypoxia Reagent (Cat Nos. I14833, I14834)?

    The difference between these products is in the final working concentration of the reagent and incubation time. For flow cytometry applications, we recommend using a final concentration in the range of 0.5 to 1 µM with an incubation time ranging from 2 to 3 hrs. For imaging, we recommend a final concentration in the range of 1 to 10 µM with an incubation time ranging from 30 mins to 1 hr. For both applications, one should optimize the final working concentration and incubation time.

    Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

    Is Hypoxia Green Reagent for Flow Cytometry (Cat No. H20035) the same reagent as the Image-iT Green Hypoxia Reagent (Cat Nos. I14833, I14834)?

    The Hypoxia Green Reagent for Flow Cytometry and the Image-iT Green Hypoxia Reagent are the same reagent, formulated differently for flow cytometry and imaging use, respectively.

    Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

    May I include a total cell stain (to stain all cells) along with Image-iT Hypoxia Reagent?

    You may use cell-permeable nuclear counterstains or surface labels, but avoid any general cytoplasmic stains that may interfere with the emission of the hypoxia reagents.

    Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

    Citations & References (1)

    Citations & References
    Abstract
    Restricted exchange microenvironments for cell culture.
    Authors:Hoh JH, Werbin JL, Heinz WF,
    Journal:Biotechniques
    PubMed ID:29570441
    'Metabolite diffusion in tissues produces gradients and heterogeneous microenvironments that are not captured in standard 2D cell culture models. Here we describe restricted exchange environment chambers (REECs) in which diffusive gradients are formed and manipulated on length scales approximating those found in vivo. In REECs, cells are grown in 2D ... More