Isolectin GS-IB4 From Griffonia simplicifolia, Alexa Fluor™ 488 Conjugate
Isolectin GS-IB4 is a 114,000-dalton glycoprotein that is part of a family of five tetrameric type I isolectins (IA4, IA3B,Read more
Have Questions?
Catalog Number
Quantity
I21411
500 μg
Catalog number I21411
Price (USD)
544.65
Online Exclusive
562.00
Save 17.35 (3%)
Each
Add to cart
Quantity:
500 μg
Price (USD)
544.65
Online Exclusive
562.00
Save 17.35 (3%)
Each
Add to cart
Isolectin GS-IB4 is a 114,000-dalton glycoprotein that is part of a family of five tetrameric type I isolectins (IA4, IA3B, IA2B2, IAB3, and IB4) isolated from the seeds of the tropical African legume Griffonia simplicifolia. The A subunit prefers N-acetyl-D-galactosamine end groups while the B subunit is selective for terminal a-D-galactosyl residues. The green fluorescent Alexa Fluor™ 488 isolectin GS-IB4 conjugate can be used in fluorescence microscopy or flow cytometry.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Label TypeAlexa Fluor Dyes
Product LineAlexa Fluor™
Protein SubtypeLectins
Quantity500 μg
Shipping ConditionRoom Temperature
SourceAfrica Shrub legume
ConjugateAlexa Fluor 488
FormLyophilized
Unit SizeEach
Contents & Storage
Store in freezer (-5 to -30°C) and protect from light.
Frequently asked questions (FAQs)
How do I know which tracer to choose for my experiment?
Factors to consider are size of tracer, method of delivery (injection, direct application to tissue, etc.), and if the tracer needs to be fixable. Here are some links to details about the various classes of neuronal tracers we offer and how to choose between them:
Neuronal Tracing (https://www.thermofisher.com/us/en/home/life-science/cell-analysis/cell-tracing-tracking-and-morphology/neuronal-tracing.html)
Choosing a Tracer (https://www.thermofisher.com/us/en/home/references/molecular-probes-the-handbook/fluorescent-tracers-of-cell-morphology-and-fluid-flow/choosing-a-tracer.html)
Imaging Analysis (http://assets.thermofisher.com/TFS-Assets/BID/Reference-Materials/bioprobes-50-journal.pdf)