The Image-iT™ LIVE Mitochondrial Transition Pore Assay Kit provides a proven method[REF PN36389 ,PN32679] of measuring mitochondrial permeability transition pore opening. The kit includes a dye and a quencher that are easily loaded into cells. In healthy cells, the dye, but not the quencher, enters mitochondria. The mitochondria remain brightly fluorescent until mitochondrial pore activation permits quenching of the fluorescence.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Format
Kit
Product Type
Mitochondrial Transition Pore Assay Kit
Color
Orange, Blue, Green
Shipping Condition
Room Temperature
Sub Cellular Localization
Mitochondria
Product Line
Image-iT™
For Use With (Equipment)
Fluorescence Microscope
Contents & Storage
Store in freezer (-5 to -30°C) and protect from light.