Image-iT™ LIVE Red and Green Caspase Apoptosis Detection Kits for microscopy
Invitrogen™

Image-iT™ LIVE Red and Green Caspase Apoptosis Detection Kits for microscopy

Detect apoptosis via microscopy, including fluorescence microscopy and HCS, with Image-iT LIVE Red and Green Caspase Apoptosis Detection Kits that report poly and caspase-3/7 activities.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberColorEnzymeLabel or Dye
I35104Green, Red, BluePoly CaspasesFAM-VAD-FMK, Propidium Iodide, Hoechst 33342
I35101Red, Green, BluePoly CaspasesSR-VAD-FMK, SYTOX Green, Hoechst 33342
I35102Red, Green, BlueCaspase 3/7SR-DEVD-FMK, SYTOX Green, Hoechst 33342
I35106Green, Red, BlueCaspase 3/7FAM-DEVD-FMK, Propidium Iodide, Hoechst 33342
Catalog number I35104
Price (USD)
361.00
Each
Color:
Green, Red, Blue
Enzyme:
Poly Caspases
Label or Dye:
FAM-VAD-FMK, Propidium Iodide, Hoechst 33342
Price (USD)
361.00
Each
Detect and evaluate caspase activation, membrane permeability, and cell cycle using Image-iT LIVE red and green poly and caspase-3 and-7 apoptosis detection kits for microscopy, including fluorescence microscopy and high content screening (HCS). The Image-iT LIVE red and green caspase detection kits use fluorescent inhibitor of caspases (FLICA) methodology to detect and report caspase activity, a measure of cell apoptosis. Specifically, the Image-iT LIVE red and green caspase detection kits contain the substrates SR-VAD-FMK and SR-DEVD-FMK (for red fluorescence), or FAM-VAD-FMK and FAM-DEVD-FMK (green fluorescence), for poly caspases and caspase-3/7 detection, respectively. They also include Hoechst 33342 and propidium iodide stains, enabling simultaneous evaluation of membrane permeability and cell cycle by microscopy and HCS.
The Image-iT LIVE red and green caspase detection kits for microscopy employ a novel approach to detecting active caspases: both assay kits take advantage of a fluorescent inhibitor of caspases (FLICA) methodology, which is an affinity label. This reagent associates a fluoromethyl ketone (FMK) moiety, which can react covalently with a cysteine, with a caspase-specific amino acid sequence. Different amino acid recognition sequences are used to detect different caspases: valine-alanine-aspartic acid (VAD) for poly-caspases (including caspase-1, -3, -4, -5, -6, -7, -8, and -9), and aspartic acid-glutamic acid-valine-aspartic acid (DEVD) for caspase 3/7. A sulforhodamine (SR) group or carboxyfluorescein (FAM) group is attached as a reporter.

The FLICA reagent is thought to interact with the enzymatic reactive center of an activated caspase via the recognition sequence, and then to attach covalently through the FMK moiety. The FLICA inhibitor is cell permeant and noncytotoxic. Unbound FLICA molecules diffuse out of the cell and are washed away; the remaining red or green fluorescent signal is a direct measure of the amount of active caspase present at the time the inhibitor was added. The approximate excitation and emission peaks of the FLICA, propidium iodide, and Hoechst 33342 reagents are 488⁄ nm/530 nm, 535⁄ nm/6617 nm, and 350 nm/⁄461 nm, respectively. The detection reagents included within the Image-iT LIVE Red Poly Caspases, Red Caspase-3/7, Green Poly Caspases, and Green Caspase-3/7 Detection kits are SR-VAD-FMK, SR-DEVD-FMK, FAM-VAD-FMK, and FAM-DEVD-FMK, respectively.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
ColorGreen, Red, Blue
DescriptionImage-iT LIVE Green Poly Caspases Detection Kit
EnzymePoly Caspases
Excitation/EmissionFAM-VAD-FMK: 488/530
propidium iodide (PI): 535/617
Hoechst 33342: 350/461
For Use With (Equipment)Fluorescence Microscope
Label TypeOther Label(s) or Dye(s)
Label or DyeFAM-VAD-FMK, Propidium Iodide, Hoechst 33342
No. of Reactions25 tests (labeling volumes of 300μL)
Product LineImage-iT
Product TypeCaspase Assay Kit
Quantity1 kit
Shipping ConditionRoom Temperature
Detection MethodFluorescence
FormatSlide
Unit SizeEach
Contents & Storage
Store in refrigerator (2–8°C) and protect from light.

Citations & References (16)

Citations & References
Abstract
Multiparametric evaluation of apoptosis: effects of standard cytotoxic agents and the cyanoguanidine CHS 828.
Authors:Lövborg H, Nygren P, Larsson R
Journal:Mol Cancer Ther
PubMed ID:15141009
'A multiparametric high-content screening assay for measurement of apoptosis was developed. HeLa cells and lymphoma U-937 cells were exposed to cytotoxic drugs in flat-bottomed optical microtiter plates. After incubation, the DNA-binding dye Hoechst 33342, fluorescein-tagged probes that covalently bind active caspases and chloromethyl-X-rosamine to detect mitochondrial membrane potential (MMP) were ... More
MiR-351 transiently increases during muscle regeneration and promotes progenitor cell proliferation and survival upon differentiation.
Authors:Chen Y, Melton DW, Gelfond JA, McManus LM, Shireman PK,
Journal:Physiol Genomics
PubMed ID:22968638
MicroRNAs (miRNAs) regulate many biological processes including muscle development. However, little is known regarding miRNA regulation of muscle regeneration. Murine tibialis anterior muscle was evaluated after cardiotoxin-induced injury and used for global miRNA expression analysis. From day 1 through day 21 following injury, 298 miRNAs were significantly changed at least ... More
Streptococcus pneumoniae evades human dendritic cell surveillance by pneumolysin expression.
Authors:Littmann M, Albiger B, Frentzen A, Normark S, Henriques-Normark B, Plant L,
Journal:EMBO Mol Med
PubMed ID:20049723
Dendritic cells (DCs) protect the respiratory epithelium via induction of innate immune responses and priming of naïve T cells during the initiation of adaptive immunity. Streptococcus pneumoniae, a commonly carried asymptomatic member of the human nasopharyngeal microflora, can cause invasive and inflammatory diseases and the cholesterol-dependent cytotoxin pneumolysin is a ... More
Marked inhibition of growth and invasive parameters of head and neck squamous carcinoma FaDu by a nutrient mixture.
Authors:Roomi MW, Roomi NW, Kalinovsky T, Rath M, Niedzwiecki A,
Journal:Integr Cancer Ther
PubMed ID:19679626
Head and neck squamous cell carcinomas (HNSCCs) are known for their aggressive growth and propensity to metastasize. The authors investigated the effect of a novel nutrient mixture (NM) containing ascorbic acid, lysine, proline, and green tea extract on human HNSCC cell line FaDu in vivo and in vitro. Athymic male ... More
The Effect of Axon Resealing on Retrograde Neuronal Death after Spinal Cord Injury in Lamprey.
Authors:
Journal:Brain Sci
PubMed ID:29661988