Informational:Like our original iBlot Dry Blotting System? Then you'll love our iBlot 3 Dry Blotting System which enables twice the throughput and features optional built-in cooling for better consistency. Learn more about the benefits of the iBlot 3 here.
Invitrogen™

iBlot™ Transfer Stack, nitrocellulose, mini

Catalog number: IB301002
Have Questions?
Invitrogen™

iBlot™ Transfer Stack, nitrocellulose, mini

Catalog number: IB301002
Have Questions?
Catalog Number
IB301002
Unit Size
10 sets
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
3 x 10 sets
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
IB30100210 sets
-
IB3010323 x 10 sets
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
iBlot™ Transfer Stacks are used to transfer proteins using the iBlot™ Gel Transfer Device. The iBlot™ Transfer Stacks are disposable stacks that come in three varieties for different applications. iBlot™ Gel Transfer Stacks have integrated PVDF or nitrocellulose transfer membranes to perform dry blotting of proteins. iBlot™ DNA Transfer Stacks have an integrated nylon transfer membrane to perform dry blotting of DNA. iBlot™ Western Detection Stacks are used to perform western detection on previously blotted nitrocellulose or PVDF membranes. Each iBlot™ Transfer Stack contains a copper electrode and appropriate cathode and anode buffers in the gel matrix to allow fast, reliable transfer of proteins or DNA.

Nitrocellulose membrane
The nitrocellulose membrane (0.2 μm pore size) is composed of 100% pure nitrocellulose to provide high-quality transfer. The membrane is compatible with commonly used detection methods such as staining, immunodetection, fluorescence, or radiolabeling. Proteins bind to the membrane due to hydrophobic and electrostatic interactions. The protein binding capacity is 209 μg/cm2.

Transfer stack sizes
The regular size (13 cm x 8.3 cm) is used for all Novex™ midi-gel types (or equivalent) and E-PAGE™ 48 or 96 Gels. The mini size (8 cm x 8 cm) is used for all Novex™ mini-gels (or equivalent).
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Length (Metric)
8 cm
Width (Metric)
8 cm
Format
Transfer Stack
Material
Nitrocellulose
Pore Size
0.2 μm
Shipping Condition
Room Temperature
Dimensions (LxW)
Mini, 8 cm x 8 cm
Product Line
iBlot
Sufficient For
10 Blots
Green Features
Sustainable disposal

Contents & Storage

10 sets of transfer stacks in 1 box. Store at room temperature.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text