iBlot™ Transfer Stack, nitrocellulose, regular size
Like our original iBlot Dry Blotting System? Then you'll love our iBlot 3 Dry Blotting System which enables twice the throughput and features optional built-in cooling for better consistency. Learn more about the benefits of the iBlot 3 here.
Invitrogen™
iBlot™ Transfer Stack, nitrocellulose, regular size
The iBlot™ Transfer Stacks are used to transfer proteins using the iBlot™ Gel Transfer Device. The iBlot™ Transfer Stacks areRead more
Have Questions?
Change view
Catalog Number
Dimensions (LxW)
Quantity
IB301031
Regular, 13.5 cm x 8 cm
3 x 10 sets
IB301001
Regular, 13.5 cm x 8 cm
10 sets
Catalog number IB301031
Price (USD)/ 3 x 10 sets
736.65
Online exclusive
788.00
Save 51.35 (7%)
-
Add to cart
Dimensions (LxW):
Regular, 13.5 cm x 8 cm
Quantity:
3 x 10 sets
Price (USD)/ 3 x 10 sets
736.65
Online exclusive
788.00
Save 51.35 (7%)
Add to cart
iBlot™ Transfer Stack, nitrocellulose, regular size
Catalog numberIB301031
Price (USD)/ 3 x 10 sets
736.65
Online exclusive
788.00
Save 51.35 (7%)
-
Add to cart
The iBlot™ Transfer Stacks are used to transfer proteins using the iBlot™ Gel Transfer Device. The iBlot™ Transfer Stacks are disposable stacks that come in three varieties for different applications: iBlot™ Gel Transfer Stacks have integrated PVDF or nitrocellulose transfer membranes to perform dry blotting of proteins, iBlot™ DNA Transfer Stacks have an integrated nylon transfer membrane to perform dry blotting of DNA, and iBlot™ Western Detection Stacks are used to perform Western detection on previously blotted nitrocellulose or PVDF membranes. Each iBlot™ Transfer Stack contains a copper electrode and appropriate cathode and anode buffers in the gel matrix to allow fast, reliable transfer of proteins or DNA.
Nitrocellulose membrane The nitrocellulose membrane (0.2 μm pore size) included in this stack is composed of 100% pure nitrocellulose to provide high-quality transfer. The membrane is compatible with commonly used detection methods such as staining, immunodetection, fluorescence, or radiolabeling. The proteins bind to the membrane due to hydrophobic and electrostatic interactions. The protein binding capacity is 209 μg/cm2.
Transfer stack sizes The regular size (13 cm x 8.3 cm) is used for all Novex™ midi-gel types (or equivalent) and E-PAGE™ 48 or 96 Gels. The mini size (8 cm x 8 cm) is used for all Novex™ mini-gels (or equivalent).