iBlot™ 3 Transfer Stacks, mini, PVDF
iBlot™ 3 Transfer Stacks, mini, PVDF
iBlot™ 3 Transfer Stacks, mini, PVDF
iBlot™ 3 Transfer Stacks, mini, PVDF
iBlot™ 3 Transfer Stacks, mini, PVDF
iBlot™ 3 Transfer Stacks, mini, PVDF
Invitrogen™

iBlot™ 3 Transfer Stacks, mini, PVDF

Green features
iBlot 3 Transfer Stacks are used with the iBlot 3 Western Blot Transfer Device for the transfer of proteins fromRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
IB3400210 Stacks
IB34002X33 x 10 Stacks
Catalog number IB34002
Price (USD)/ Each
250.65
Online Exclusive
261.00
Save 10.35 (4%)
Add to cart
Quantity:
10 Stacks
Price (USD)/ Each
250.65
Online Exclusive
261.00
Save 10.35 (4%)
Add to cart

iBlot 3 Transfer Stacks are used with the iBlot 3 Western Blot Transfer Device for the transfer of proteins from an SDS-PAGE gel to a membrane support when performing western blot experiments. iBlot 3 Transfer Stacks contain copper-fiber electrode sheets, optimized cathode and anode solid buffer gels, and a preconditioned PVDF transfer membrane. They are compatible with common commercially available protein gels, including Bis-Tris, Tris-Glycine, Tris-Acetate, and Tricine chemistries. This mini-size stack (8 x 8 cm) can be used with one mini-size gel.

The iBlot 3 Western Blot Transfer System (iBlot 3 Transfer Stacks plus iBlot3 Transfer Device) has been optimized to provide protein transfers in 3–8 minutes that produce higher quality and more reliable western blot result compared to many other transfer methods.

PVDF membrane

The PVDF membrane (0.2-μm pore size, low fluorescence) in this stack has higher binding capacity than nitrocellulose. It is pre-activated and ready for use without any pretreatment with alcohols. The membrane is compatible with commonly used detection methods such as staining, immunodetection, fluorescence, and radiolabeling. Proteins bind to the membrane due to hydrophobic interactions. The protein-binding capacity is 240 μg/cm2.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Green FeaturesLess waste, sustainable packaging, sustainable disposal
Quantity10 Stacks
Dimensions (LxW)8 x 8 cm (Mini)
FormatTransfer Stack
Length (Metric)8 cm
MaterialPVDF
Pore Size0.2 μm
Product LineiBlot
Sufficient For10 Blots
Width (Metric)8 cm
Unit SizeEach
Contents & Storage
Store at room temperature.

Save up to 24% off on Protein Analysis products

Share catalog number, name or link