Search by lot number or partial lot number
Get significant increases in resolution and throughput using the world's first modular ion chromatography (IC) system capable of operation up to 5000 psi. The Thermo Scientific™ Dionex™ ICS-6000 Standard Bore and Microbore System can operate continuously at up to 5000 psi when configured as a Reagent-Free™ IC (RFIC™) system. Get higher resolution using smaller particle size (4μm) columns, faster run times without sacrificing needed resolution using higher flow rates and 150mm columns, or higher resolution with standard flow rates and 250mm columns. The ability to configure this flexible, modular system for an unmatched variety of applications.
Type | Single Channel Standard Bore RFIC System |
Includes | 1x SP Pump, Isocratic with Degas 1x DC (SB) with Dual Temperature Zone, One Injection Valve 1x EG Module 1x EG Degas Unit (SB/MB) 1x CD Detector (with Cell) 1x EO Eluent Organizer Tray with two 2L bottles 1x IC PEEK Viper Fitting Kit for Dionex ICS-6000 with CD (Analytical) 1x Consumable Device Monitor 1x Tablet Kit (with arm, holder, and connectivity kit) |
For Use With (Application) | Ion Chromatography |
Description | Dionex ICS-6000 Single Channel Standard Bore RFIC System |
Unit Size | Each |
Type | Single Channel Standard Bore IC System |
Includes | 1 x SP Pump, Isocratic with Degas<br /> 1 x DC (SB) with Dual Temperature Zone, One Injection Valve<br /> 1 x CD Detector (with Cell)<br /> 1 x EO Eluent Organizer Tray with two 2 Liter bottles<br /> 1 x IC PEEK Viper Fitting Kit for Dionex ICS-6000 with CD (Analytical)<br /> 1 x Consumable Device Monitor<br /> 1 x Tablet Kit (with arm, holder, and connectivity kit) |
For Use With (Application) | Ion Chromatography |
Description | Dionex ICS-6000 Single Channel Standard Bore IC System |
Unit Size | Each |
Type | Single Channel Gradient Standard Bore IC System |
Includes | 1x SP Pump, Quaternary with Degas 1x DC (SB) with Dual Temperature Zone, One Injection Valve 1x CD Detector (with Cell) 1x EO Eluent Organizer Tray with four 2 Liter bottles 1x IC PEEK Viper Fitting Kit for Dionex ICS-6000 with CD (Analytical) 1x Consumable Device Monitor 1x Tablet Kit (with arm, holder, and connectivity kit) |
For Use With (Application) | Ion Chromatography |
Description | Dionex ICS-6000 Single Channel Gradient Standard Bore IC System |
Unit Size | Each |
Type | Single Channel Microbore RFIC System |
Includes | 1x SP Pump, Isocratic with Degas 1x DC (MB) with Dual Temperature Zone, One Injection Valve 1x EG Module 1x EG Degas Unit (SB/MB) 1x CD Detector (with Cell) 1x EO Eluent Organizer Tray with two 2L bottles 1x IC PEEK Viper Fitting Kit for Dionex ICS-6000 with CD (Analytical) 1x Consumable Device Monitor 1x Tablet Kit (with arm, holder, and connectivity kit) |
For Use With (Application) | Ion Chromatography |
Description | Dionex ICS-6000 Single Channel Microbore RFIC System |
Unit Size | Each |
Type | Dual Channel Standard Bore RFIC System |
Includes | 1x DP Pump, Iso/Iso (Analytical) with Degas 1x DC (SB) with Dual Temperature Zone, Two Injection Valves 1x EG Module 2x EG degas unit (SB/MB) 2x CD Detector (with Cell) 1x EO Eluent Organizer Tray with four 2L bottles 2x IC PEEK Viper Fitting Kit for Dionex ICS-6000 with CD (Analytical) 1x Consumable Device Monitor 1x Tablet Kit (with arm, holder, and connectivity kit) |
For Use With (Application) | Ion Chromatography |
Description | Dionex ICS-6000 Dual Channel Standard Bore RFIC System |
Unit Size | Each |
Type | Dual Channel Microbore RFIC System |
Includes | 1x DP Pump, Iso/Iso with Degas 1x DC (MB) with Dual Temperature Zone, Two Injection Valves 1x EG Module 2x EG degas unit (SB/MB) 2x CD Detector (with Cell) 1x EO Eluent Organizer Tray with four 2L bottles 2x IC PEEK Viper Fitting Kit for Dionex ICS-6000 with CD (Analytical) 1x Consumable Device Monitor 1x Tablet Kit (with arm, holder, and connectivity kit) |
For Use With (Application) | Ion Chromatography |
Description | Dionex ICS-6000 Dual Channel Microbore RFIC System |
Unit Size | Each |
Type | 2D-IC Standard Bore-Microbore System |
Includes | 1x DP Pump, Iso/Iso with Degas 1x DC with Dual Temperature Zone (SB), One Injection Valve 1x 6-port valve upgrade kit (MB) 1x Microbore Tubing Kit 1x Microbore Temperature Kit 2x CD Detector (with Cell) 1x EG Module 2x Degas Unit for EG (SB/MB) 1x EO Eluent Organizer Tray with four 2L bottles 1x Tablet Kit (with arm, holder, and connectivity kit) |
For Use With (Application) | Ion Chromatography |
Description | Dionex ICS-6000 2D-IC Standard Bore-Microbore System |
Unit Size | Each |
Type | Carbohydrate Analysis Standard Bore HPIC System |
Includes | 1x SP Pump, Quaternary with Degas 1x DC with Dual Temperature Zone, One Injection Valve 1x ED Detector 1x ED Detector Cell 1x ED Detector reference electrode (Ag/AgCl) 1x Gold Disposable Electrode 1x EO Eluent Organizer Trays with four 2L bottles 1x EO Regulator Accessory and Stand 1x IC PEEK Viper Fitting Kit for Dionex ICS-6000 with ED (Analytical) 1x Tablet Kit (with arm, holder, and connectivity kit) |
For Use With (Application) | Carbohydrate Analysis, Ion Chromatography |
Description | Dionex ICS-6000 Carbohydrate Analysis Standard Bore System |
Unit Size | Each |
Type | BioIC System with ED |
Includes | 1x DP Pump, Quat/Quat (Analytical) with Degas 1x DC with Dual Temperature Zone (SB), One Injection Valve 1x 6-port Valve Upgrade Kit 1x Microbore Tubing Kit 1x Microbore Temperature Kit 2x ED Detectors 2x ED Detector Cells 2x ED Detector Reference Electrodes (Ag/AgCl) 1x Gold Disposable Electrode (Au on PTFE) 1x Gold Disposable Electrode (AAA-certified) 1x Tablet Kit (with arm, holder, and connectivity kit) |
For Use With (Application) | Carbohydrates (Capillary) and AAA-Direct (Microbore), Ion Chromatography |
Description | Dionex ICS-6000 BioIC System with ED for Carbohydrates (Standard Bore) and AAA-Direct (Microbore) System |
Unit Size | Each |
Catalog Number | Specifications | Unit Size | Description | Price (USD) | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
ICS6000-009 | Each | Dionex ICS-6000 Single Channel Standard Bore RFIC System | Request A Quote | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
ICS6000-007 | Each | Dionex ICS-6000 Single Channel Standard Bore IC System | Request A Quote | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
ICS6000-008 | Each | Dionex ICS-6000 Single Channel Gradient Standard Bore IC System | Request A Quote | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
ICS6000-010 | Each | Dionex ICS-6000 Single Channel Microbore RFIC System | Request A Quote | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
ICS6000-011 | Each | Dionex ICS-6000 Dual Channel Standard Bore RFIC System | Request A Quote | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
ICS6000-012 | Each | Dionex ICS-6000 Dual Channel Microbore RFIC System | Request A Quote | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
ICS6000-013 | Each | Dionex ICS-6000 2D-IC Standard Bore-Microbore System | Request A Quote | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
ICS6000-014 | Each | Dionex ICS-6000 Carbohydrate Analysis Standard Bore System | Request A Quote | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
ICS6000-015 | Each | Dionex ICS-6000 BioIC System with ED for Carbohydrates (Standard Bore) and AAA-Direct (Microbore) System | Request A Quote | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|
Broad variety of modules and related products
Flexible choice of detectors