Thermo Scientific™
Watson LIMS™ Software
Catalog number: INF-21000
Have Questions?
Thermo Scientific™
Watson LIMS™ Software
Catalog number: INF-21000
Have Questions?
Manage bioanalysis studies from project initiation through study closeout with an industry leading bioanalysis solution, Thermo Scientific™ Watson LIMS™ software. Watson LIMS software was designed with a deep understanding of small and large molecule bioanalytical workflows. The software helps to enable compliance with industry regulations such as 21 CFR Part 58 for small and large molecule bioanalysis.  Study coordinators, scientists, and technical staff are all able to move logically through each step in their process, simplifying bioanalytical support for nonclinical and clinical studies.
Learn more
Catalog Number
INF-21000
Unit Size
Each
Type
Watson LIMS Software
Price (USD)
Full specifications
TypeWatson LIMS Software
Unit SizeEach
Showing 1 of 1
Catalog NumberSpecificationsUnit SizeTypePrice (USD)
INF-21000Full specifications
EachWatson LIMS SoftwareRequest A Quote
TypeWatson LIMS Software
Unit SizeEach
Showing 1 of 1
Thermo Scientific™ Watson LIMS™ software is specifically designed and built to support the bioanalytical workflow, allowing users to:
  • Manage both small and large molecule bioanalysis workflows in a single solution
  • Harmonize laboratory processes across all sites
  • Ease exchange of information between sponsors and contract labs
  • Increase throughput by automating data handling and reporting tasks
  • Promote compliance with standards such as 21 CFR Part 58 throughout the workflow
  • Implement quantitative concentration assays or qualitative immunogenicity assays
  • Track study sample history from receipt to disposal
  • Comply with SOPs and reduce the risk of non-compliance