Applied Biosystems™

VetMAX™ Swine Influenza A-09 Kit

Catalog number: INFAPSWINE
Have Questions?
Applied Biosystems™

VetMAX™ Swine Influenza A-09 Kit

Catalog number: INFAPSWINE
Have Questions?
Catalog Number
INFAPSWINE
Unit Size
100 tests
Price (USD)
Catalog NumberUnit SizePrice (USD)
INFAPSWINE100 tests
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Note: The name of this product has changed. It was previously called LSI VetMAX SWINE Influenza A/H1N1/2009.

The LSI VetMAX™ Swine Influenza A (A/H1N1/2009) Kit is a molecular diagnostic tool for influenza type A virus. The kit serves to specifically detect a highly conserved region of the M gene which is specific to all influenza A viruses, including the pandemic Influenza A/H1N1/2009 strain, by real-time reverse transcriptase PCR. It does not detect other strains of influenza (types B and C).

Each RNA sample is analyzed in a single well; the same well is used to specifically detect the viral RNA of influenza type A and an Internal Positive Control (IPC). A positive IPC signifies both successful extraction and the absence of PCR inhibitors in the sample.

This kit can be used on viral RNA extracted from nasal swabs, organs, bronchoalveolar lavage, amniotic fluid, and cell culture supernatant.

Specifications

Detection Method
Primer-probe
GC-Rich PCR Performance
High
Disease
Swine Flu
Product Line
LSI VetMAX™
Product Type
PCR Reagents and Detection Kit for Animal Health
Species
Swine/Porcine
Target Organism Class
Swine Influenza Virus

Contents & Storage

Contains components for detecting influenza type A and an internal positive control (IPC).
Upon receipt, the kit should be stored at -5°C to -30°C before and after opening.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text