3D tomography software

Thermo Scientific Inspect 3D Software provides a user-friendly solution for alignment of tomographic image series (obtained with transmission electron microscopy or scanning transmission electron microscopy, TEM or STEM) along with energy dispersive X-ray spectroscopy (EDS) maps, producing a 3D reconstruction of the data. Inspect 3D Software also offers basic visualization functionality. All Thermo Scientific TEM, STEM and EDS tomography software are fully compatible with Inspects 3D Software.

Inspect 3D Software begins with a coarse X-Y alignment based on image cross-correlation. Alignment can be further refined with automated bead tracking or patch tracking procedures. Bead tracking traces multiple round particles on the specimen throughout the tilt series, correcting rotation and magnification changes for each image. Patch tracking traces multiple areas of the image throughout the tilt series and then combines the results.

Inspect 3D Software offers two classical algorithms for 3D volume reconstruction: weighted back projection (WBP) and simultaneous iterative reconstruction technique (SIRT). Additionally, Inspect 3D Software offers easy EDS-tomography reconstruction with simultaneous processing of STEM and EDS files, called HAADF-EDS bimodal tomography (HEBT), leading to enhanced elemental reconstruction.

Iterative reconstruction techniques offered with Inspect 3D Software include;

  • Conjugate gradient least squares (CGLS)
  • Expectation maximization (EM), in which elongation artifacts are significantly reduced when compared to SIRT reconstruction
  • Simultaneous algebraic reconstruction technique (SART)

The resulting reconstructed volumes can be directly imported into Thermo Scientific Avizo and Amira Software for further visualization and analysis.

Key Features

  • Cross-correlation requires no markers as the process is enhanced by the use of a large variety of filters and image processing tools.
  • Feature tracking can utilize added artificial features like gold markers, or recognizable intrinsic specimen features, as a basis for image alignment.
  • Bead cloaking removes gold beads from the image data after they have been used for alignment, so they do not show in the reconstruction. 
  • The algebraic reconstruction technique iteratively compares projections of the reconstruction to the original projections and corrects the reconstruction to minimize differences.
  • The simultaneous iterative reconstruction technique preserves full spatial information and can enhance the signal to noise ratio of the reconstruction. 
 
Tomography post-alignment and reconstruction module
Tomography post-alignment and reconstruction module
Inspect 3D reconstructed 3D volume screen capture
Inspect 3D reconstructed 3D volume screen capture
 

Applications

Infectious_Disease_Research_App_w274x180

Pesquisa de doenças infecciosas
 

As técnicas da crio-EM (microscopia crioeletrônica) permitem observações em várias escalas de estruturas biológicas 3D em seus estados quase nativos, informando um desenvolvimento mais rápido e eficiente da terapêutica.

Plant_Biology_Research_Application_274x180_144_DPI

Pesquisa de biologia vegetal
 

A pesquisa fundamental da biologia vegetal é possível devido à microscopia crioeletrônica. Ela fornece informações sobre proteínas (com análise de partículas únicas), sobre seu contexto celular (com tomografia), até a estrutura geral da planta (análise de volume grande).

Pathology_Pancreas_274x180

Pesquisa de patologia
 

A microscopia eletrônica de transmissão (TEM) é usada quando a natureza da doença não pode ser estabelecida por métodos alternativos. Com a geração de imagens nanobiológicas, a TEM fornece informações precisas e confiáveis para determinadas patologias.

Process Control_Thumb_274x180_144DPI

Controle de processo
 

A indústria moderna exige alta produtividade com qualidade superior, um equilíbrio mantido por meio de um controle de processo robusto. As ferramentas SEM e TEM com software de automação dedicado proporcionam informações rápidas e em várias escalas para monitoramento e aprimoramento de processos.

 

Quality Control_Thumb_274x180_144DPI

Controle de qualidade
 

O controle de qualidade e a garantia de qualidade são essenciais na indústria moderna. Oferecemos uma gama de ferramentas de microscopia eletrônica e espectroscopia para análises multidimensionais e multimodais de defeitos, permitindo que você tome decisões confiáveis e informadas para controle e melhoria de processos.

 

Fundamental Materials Research_R&D_Thumb_274x180_144DPI

Pesquisa de materiais fundamentais

Novos materiais são investigados em escalas cada vez menores para o máximo controle de suas propriedades físicas e químicas. A microscopia eletrônica fornece aos pesquisadores percepções importantes sobre uma ampla variedade de características materiais em escala micro a nano.

 

pathfinding_thumb_274x180_144dpi

Desenvolvimento e localização de semicondutores

Microscopia eletrônica avançada, feixe de íon focalizado e técnicas analíticas associadas para identificar soluções viáveis e métodos de desenho para a fabricação de dispositivos semicondutores de alto desempenho.

Análise de falha de semicondutores

Análise de falha de semicondutores

Estruturas de dispositivos semicondutores cada vez mais complexas resultam em mais locais onde defeitos que induzem falhas podem se ocultar. Nossos fluxos de trabalho de última geração o ajudam a localizar e caracterizar problemas elétricos sutis que afetam o rendimento, o desempenho e a confiabilidade.

physical_characterization_thumb_274x180_144dpi

Caracterização física e química

A demanda contínua dos consumidores impulsiona a criação de dispositivos eletrônicos menores, mais rápidos e mais baratos. Sua produção depende de instrumentos e fluxos de trabalho de alta produtividade que fazem imagens, analisam e caracterizam uma ampla gama de semicondutores e dispositivos de exibição.

 
 
 

Techniques

Style Sheet for Komodo Tabs

Análise de partícula única

A análise de partículas únicas (SPA) é uma técnica de microscopia crioeletrônica que permite a caracterização estrutural em resoluções quase atômicas, desvendando processos biológicos dinâmicos e a estrutura de complexos/conjuntos biomoleculares.

Saiba mais ›

Criotomografia

A tomografia crioeletrônica (crio-ET) fornece tanto informações estruturais sobre proteínas individuais como seus arranjos espaciais dentro da célula. Isso faz dela uma técnica verdadeiramente única e também explica por que o método tem um potencial imenso para a biologia celular. A tomografia crioeletrônica pode preencher a lacuna entre microscopia de luz e técnicas de resolução quase atômica, como a análise de partículas únicas.

Saiba mais ›

Caracterização de materiais 3D

O desenvolvimento de materiais muitas vezes requer caracterização 3D em várias escalas. Os instrumentos DualBeam permitem a secção em série de grandes volumes e a subsequente geração de imagens SEM nanométricas, que podem ser processadas em reconstruções 3D de alta qualidade da amostra.

Saiba mais ›

Tomografia EDS 3D

A pesquisa de materiais modernos depende cada vez mais da análise nanométrica em três dimensões. A caracterização 3D, incluindo dados de composição para contexto químico e estrutural completos, é possível com a EM 3D e a espectroscopia de raios X por energia dispersiva.

Saiba mais ›

Aquisição de imagens e análise TEM de semicondutores

Os microscópios eletrônicos de transmissão da Thermo Fisher Scientific oferecem imagens de alta resolução e análise de dispositivos semicondutores, permitindo que os fabricantes calibrem conjuntos de ferramentas, diagnostiquem mecanismos de falha e otimizem o rendimento geral do processo.

Saiba mais ›

Análise de partícula única

A análise de partículas únicas (SPA) é uma técnica de microscopia crioeletrônica que permite a caracterização estrutural em resoluções quase atômicas, desvendando processos biológicos dinâmicos e a estrutura de complexos/conjuntos biomoleculares.

Saiba mais ›

Criotomografia

A tomografia crioeletrônica (crio-ET) fornece tanto informações estruturais sobre proteínas individuais como seus arranjos espaciais dentro da célula. Isso faz dela uma técnica verdadeiramente única e também explica por que o método tem um potencial imenso para a biologia celular. A tomografia crioeletrônica pode preencher a lacuna entre microscopia de luz e técnicas de resolução quase atômica, como a análise de partículas únicas.

Saiba mais ›

Caracterização de materiais 3D

O desenvolvimento de materiais muitas vezes requer caracterização 3D em várias escalas. Os instrumentos DualBeam permitem a secção em série de grandes volumes e a subsequente geração de imagens SEM nanométricas, que podem ser processadas em reconstruções 3D de alta qualidade da amostra.

Saiba mais ›

Tomografia EDS 3D

A pesquisa de materiais modernos depende cada vez mais da análise nanométrica em três dimensões. A caracterização 3D, incluindo dados de composição para contexto químico e estrutural completos, é possível com a EM 3D e a espectroscopia de raios X por energia dispersiva.

Saiba mais ›

Aquisição de imagens e análise TEM de semicondutores

Os microscópios eletrônicos de transmissão da Thermo Fisher Scientific oferecem imagens de alta resolução e análise de dispositivos semicondutores, permitindo que os fabricantes calibrem conjuntos de ferramentas, diagnostiquem mecanismos de falha e otimizem o rendimento geral do processo.

Saiba mais ›

Style Sheet to change H2 style to p with em-h2-header class

Contact us

Serviços de microscopia eletrônica

Para garantir o desempenho ideal do sistema, fornecemos acesso a uma rede de especialistas em serviços de campo, suporte técnico e peças de reposição certificadas.

Saiba mais ›

Folha de estilo para Rodapé de suporte e serviço
Folha de estilo para Fontes
Folha de estilo para Cartões