Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Thermo Scientific™ UltiMate™ 3000 Rapid Separation systems combine unique hardware features and the ability to use small particle columns for fast separations with excellent resolution. All system components are coordinated for high performance. Available in biocompatible and non-biocompatible setups and featuring unique technological innovations like dual-gradient capabilities, the UltiMate 3000 RS systems sustain even most advanced workflows.
Rapid Separation (RS) systems provide ultra-fast separations with ultra-high resolution in an exceptionally versatile HPLC/UHPLC system for the rigorous demands of large and small molecule analysis. These systems operate at pressures up to 1000 bar.
Biocompatible Rapid Separation (BioRS) systems provide UHPLC performance for proteins, peptides, nucleic acids, and biotherapeutics. These systems are 100% corrosion free for use with high salt or extreme pH conditions. They operate with column pressures up to 1000 bar.
Type | Rapid Separation System |
Description | UltiMate 3000 Rapid Separation Dual System |
Solvent Channels | 6 or 2 x 3 |
Max. Pressure | 100 MPa |
Detector Data Collection Rate | 200 Hz Max. |
Column Temperature Range | 5°C to 110°C |
Max. Flow Rate | 8mL/min |
Unit Size | Each |
Type | Biocompatible Rapid Separation System |
Description | UltiMate 3000 Biocompatible Rapid Separation Dual System |
Solvent Channels | 6 or 2 x 3 |
Max. Pressure | 100 MPa |
Detector Data Collection Rate | 200 Hz Max. |
Column Temperature Range | 5°C to 110°C |
Max. Flow Rate | 8mL/min |
Unit Size | Each |
Type | Biocompatible Rapid Separation System |
Description | UltiMate 3000 Biocompatible Rapid Separation System |
Solvent Channels | 2 or 4 |
Max. Pressure | 100 MPa |
Detector Data Collection Rate | 200 Hz Max. |
Column Temperature Range | 5°C to 110°C |
Max. Flow Rate | 8mL/min |
Unit Size | Each |
Type | Rapid Separation System |
Description | UltiMate 3000 Rapid Separation Quaternary System |
Solvent Channels | 4 |
Max. Pressure | 100 MPa |
Detector Data Collection Rate | 200 Hz Max. |
Column Temperature Range | 5°C to 110°C |
Max. Flow Rate | 8mL/min |
Unit Size | Each |
Type | Rapid Separation System |
Description | UltiMate 3000 Rapid Separation Binary System |
Solvent Channels | 2 or 4 |
Max. Pressure | 100 MPa |
Detector Data Collection Rate | 200 Hz Max. |
Column Temperature Range | 5°C to 110°C |
Max. Flow Rate | 8mL/min |
Unit Size | Each |
Catalog Number | Specifications | Unit Size | Description | Solvent Channels | Max. Pressure | Detector Data Collection Rate | Price (USD) | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
IQLAAAGABHFAPBMBFE | Each | UltiMate 3000 Rapid Separation Dual System | 6 or 2 x 3 | 100 MPa | 200 Hz Max. | Request A Quote | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
IQLAAAGABHFAPBMBFF | Each | UltiMate 3000 Biocompatible Rapid Separation Dual System | 6 or 2 x 3 | 100 MPa | 200 Hz Max. | Request A Quote | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
IQLAAAGADGFAMIMBCO | Each | UltiMate 3000 Biocompatible Rapid Separation System | 2 or 4 | 100 MPa | 200 Hz Max. | Request A Quote | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
IQLAAAGABHFAPBMBFA | Each | UltiMate 3000 Rapid Separation Quaternary System | 4 | 100 MPa | 200 Hz Max. | Request A Quote | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
IQLAAAGABHFAPBMBEZ | Each | UltiMate 3000 Rapid Separation Binary System | 2 or 4 | 100 MPa | 200 Hz Max. | Request A Quote | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|
Basic HPLC systems
UltiMate 3000 Basic Automated System
Enjoy cost effective, automated HPLC and UHPLC. Ideal for teaching or moderate throughput applications. Includes an isocratic or quaternary pump and integrated autosampler and heated column compartment.
UltiMate 3000 Basic Manual System
Enjoy cost effective access to HPLC and UHPLC separations. System features an isocratic pump and manual sample injection in a simple, expandable system where modularity makes anything possible.
Ensuring Excellent Chromatography
Chromeleon CDS Software Makes Operation Easy
Choose the Right Configuration for You
Basic HPLC Systems
Basic Automated
Basic Manual