Search by lot number or partial lot number
Quantify persistent organic pollutants (POPs), routinely in a regulated environment, using Thermo Scientific™ TargetQuan 3 software. Uniquely tailored for workflow-driven POP quantification, the software provides comprehensive processing of MS, MS/MS and high-resolution MS data on a single software platform. TargetQuan 3 software supports LC-MS and GC-MS data, including high-resolution GC-MS data.
Type | Software |
Description | TargetQuan 3 Software |
For Use With (Equipment) | LC-MS and GC-MS Systems |
Unit Size | Each |
Catalog Number | Specifications | Unit Size | Price (USD) | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
IQLAAAGABTFAIAMAQE | Each | Request A Quote | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|