Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
For Use With (Equipment) | Thermo Scientific Antaris II FT-NIR spectrometer |
Type | MultiPro Autosampler for Antaris NIR |
Sample Type | Tablet, Caplets, and Soft Gels |
Unit Size | Each |
Catalog Number | Specifications | Unit Size | For Use With (Equipment) | Sample Type | Type | Price (USD) | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
IQLAADGABCFADMMAJF | Each | Thermo Scientific Antaris II FT-NIR spectrometer | Tablet, Caplets, and Soft Gels | MultiPro Autosampler for Antaris NIR | Request A Quote | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|
Features
Recommended for: