Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Simultaneously analyze more than 100 gas species using the Thermo Scientific™ Antaris™ IGS Gas Analyzer. Developed with input from industry market leaders to solve specific gas analysis needs, this multi-component gas analyzer provides research-grade performance, speed and sensitivity in a package designed specifically for use in plant and process environments.
Product Type | Antaris Industrial Gas System (IGS) |
Components Measured | Up to 25 components measure simultaneously |
Description | Multi-component gas system capable of simultaneously analyzing over 100 gas species. |
Hertz | Sample rate up to 5 Hz |
Options | Single spectrum or continuous monitoring |
Sample Type | Gas cells and all standard sample compartment accessories |
Voltage | 120/240 V |
Unit Size | Each |
Catalog Number | Specifications | Unit Size | Description | Sample Type | Price (BRL) | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
IQLAADGABCFADUMAAU | Each | Multi-component gas system capable of simultaneously analyzing over 100 gas species. | Gas cells and all standard sample compartment accessories | Contact Us › | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|
The Antaris IGS Gas Analyzer Features:
Thermo Scientific™ RESULT™ Software in the Antaris IGS Gas Analyzer provides easy-to-use tools for routine analysis, method development, method transfer, operation setup and data analysis.
Recommended for: