Thermo Scientific™
Avantage Data System
Catalog number: IQLAADGACKFAKRMAVI
Have Questions?
Thermo Scientific™
Avantage Data System
Catalog number: IQLAADGACKFAKRMAVI
Have Questions?
Realize the full potential of your electron spectrometer with the Thermo Scientific™ Avantage Data System for surface analysis. This best-in-class data system integrates instrument control, data acquisition, data processing and reporting to provide precise, accurate and reliable instrument control.
Whether working in a dedicated research lab or as a tool in a multi-user environment, users of all abilities can rely on the flexibility, feature-set, and intuitive operation of the Avantage Data System to obtain the maximum information from their samples.
Catalog Number
IQLAADGACKFAKRMAVI
Unit Size
Each
Description
Avantage Data System
Price (USD)
Full specifications
DescriptionAvantage Data System
Unit SizeEach
Showing 1 of 1
Catalog NumberSpecificationsUnit SizeDescriptionPrice (USD)
IQLAADGACKFAKRMAVIFull specifications
EachAvantage Data SystemRequest A Quote
DescriptionAvantage Data System
Unit SizeEach
Showing 1 of 1
Avantage Data System Features:
  • Instrument control for all Thermo Scientific™ Surface Analysis Systems
  • Fully-flexible experiment design
  • Automated data acquisition including data processing functions and reporting
  • Comprehensive libraries, references and intelligent algorithms for data interpretation
  • Advanced data reduction tools
  • Flexible reporting functions