Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
The Thermo Scientific Nexsa G2 X-Ray Photoelectron Spectrometer (XPS) System offers fully automated, high-throughput surface analysis, delivering the data to advance research and development or to solve production problems. By integrating XPS with ion scattering spectroscopy (ISS), UV photoelectron spectroscopy (UPS), reflected electron energy loss spectroscopy (REELS), and Raman spectroscopy, it allows you to conduct true correlative analysis.
The system now includes options for sample heating and sample biasing capabilities to increase the range of possible experiments. The Nexsa G2 Surface Analysis System unlocks the potential for advances in materials science, microelectronics, nanotechnology development, and many other applications.
A new, optimized X-ray monochromator design makes it possible to select an analysis area from 10 µm to 400 µm in 5 µm steps, ensuring data is collected from the feature of interest while maximizing the signal.
The high-efficiency electron lens, hemispherical analyzer, and detector allow for superb detectability and rapid data acquisition.
Bring sample features into focus with the Nexsa G2 Surface Analysis System's patented optical viewing system and XPS SnapMap, which helps you pinpoint areas of interest quickly.
The patented dual beam flood source couples low-energy ion beams with very low energy electrons (less than 1 eV) to prevent sample charging during analysis, which eliminates the need, in most cases, for charge referencing.
Go beyond the surface with the EX06 ion source. Automated source optimization and gas handling ensure excellent performance and experimental reproducibility.
Expand your possibilities with available specialist sample holders for angle-resolved XPS, sample bias measurements, or inert transfer from a glove box.
Instrument control, data processing, and reporting are all controlled from Thermo Scientific Avantage Software. Avantage Software includes Knowledge View, a comprehensive source of tutorials and reference material that guide users through analysis.
Correlate XPS data with SEM images through the correlative imaging and surface analysis workflow using Thermo Scientific Maps Software.
Analyzer type |
|
X-ray source type |
|
X-ray spot size |
|
Depth profiling |
|
Maximum sample area |
|
Maximum sample thickness |
|
Vacuum system |
|
Optional accessories |
|
For Research Use Only. Not for use in diagnostic procedures.