Thermo Scientific™

Q Exactive™ Plus Hybrid Quadrupole-Orbitrap™ Mass Spectrometer

Catalog number: IQLAAEGAAPFALGMBDK
Have Questions?
Thermo Scientific™

Q Exactive™ Plus Hybrid Quadrupole-Orbitrap™ Mass Spectrometer

Catalog number: IQLAAEGAAPFALGMBDK
Have Questions?

Take Quanfirmation™ to the next level. The Thermo Scientific™ Q Exactive™ Hybrid Quadrupole-Orbitrap™ Mass Spectrometer introduced the revolutionary concept of Quanfirmation. The Q Exactive™ Plus Hybrid Quadrupole-Orbitrap™ Mass Spectrometer takes Quanfirmation to the next level with increased performance and new options that enhance applications from confident DMPK qual/quan screening studies to characterization of intact monoclonal antibodies.

 
Catalog Number
IQLAAEGAAPFALGMBDK
Unit Size
Each
Scan Rates
Up to 12 Hz
Mass Range
50 to 6000 m/z
Price (USD)
Full specifications
TypeQuadrupole-Orbitrap LC-MS
Dynamic Range>5000:1
Mass Range50 to 6000 m/z
Mass AccuracyInternal: <1 ppm RMS
External: <3 ppm RMS
Scan RatesUp to 12 Hz
Polarity SwitchingOne Full Cycle in <1 Sec (One Full Positive Mode Scan and One Full Negative Mode Scan (at a Resolution Setting of 35,000))
SensitivityFull MS: 500 fg Buspirone on Column S/N 100:1
SIM: 30 fg Buspirone on Column S/N 100:1
Max. ResolutionUp to 240,000 at m/z 200
Unit SizeEach
Showing 1 of 1
Catalog NumberSpecificationsUnit SizeScan RatesMass RangePrice (USD)
IQLAAEGAAPFALGMBDKFull specifications
EachUp to 12 Hz50 to 6000 m/zRequest A Quote
TypeQuadrupole-Orbitrap LC-MS
Dynamic Range>5000:1
Mass Range50 to 6000 m/z
Mass AccuracyInternal: <1 ppm RMS
External: <3 ppm RMS
Scan RatesUp to 12 Hz
Polarity SwitchingOne Full Cycle in <1 Sec (One Full Positive Mode Scan and One Full Negative Mode Scan (at a Resolution Setting of 35,000))
SensitivityFull MS: 500 fg Buspirone on Column S/N 100:1
SIM: 30 fg Buspirone on Column S/N 100:1
Max. ResolutionUp to 240,000 at m/z 200
Unit SizeEach
Showing 1 of 1

Superior Quantitation

  • Advanced quadrupole technology (AQT) improves precursor selection and transmission for more-accurate quantitation of low-abundance analytes in complex matrices
  • Sophisticated data-independent acquisition (DIA) and parallel reaction monitoring (PRM) deliver reproducible quantitation with complete qualitative confidence
  • An advanced active beam guide (AABG) reduces noise and extends maintenance intervals

Enhanced Analysis of Intact Proteins

  • Optional intact protein mode provides superior trapping of large molecules for improved analysis of intact proteins and protein complexes
  • Optional enhanced resolution ensures maximum ID confidence in top-down proteomics and lipidomics

High-Confidence Results

  • HR/AM and full-scan capabilities capture all sample data, all of the time, enabling retrospective data analysis without need to re-run samples
  • Resolving power up to 140,000 FWHM eliminates isobaric interferences, increasing confidence in results when analyzing samples in complex matrices
  • Better than 1-ppm mass accuracy in full- and all-ion fragmentation (AIF) scan modes ensures confident compound identification
  • Fast scanning at 12 Hz and spectral multiplexing suited to UHPLC applications
  • Rapid polarity switching maximizes information obtained
  • AIF and multiple dissociation techniques — in-source CID and HCD — aid in compound identification
  • Mass range to 6000 m/z enhances detection of singly charged small molecules and biomolecules
  • More than four orders of magnitude intrascan dynamic range, along with femtogram-level sensitivity, allow detection of trace-level and high-abundance compounds in the same scan

For Research Use Only. Not for use in diagnostic procedures.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text