Search Thermo Fisher Scientific
Achieve highly sensitive, ultra-precise isotopic measurements in an automated, high-throughput workflow with the Thermo Scientific™ Kiel IV Carbonate Device. This device, when coupled to a Thermo Scientific™ 253 Plus™ 10 kV isotope ratio mass spectrometer (IRMS), enables analysis of carbonate samples during paleoclimatic reconstructions involving biogenic carbonates such as foraminifera, bivalves, brachiopods, otoliths, and corals. Other applicable fields of study include analysis of growth zones of individual organisms or microfossils from drill cores.
Type | IRMS |
Description | Kiel IV Carbonate Device |
Unit Size | Each |
Catalog Number | Specifications | Unit Size | Description | Price (USD) | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
IQLAAEGAATFAETMAHW | Each | Kiel IV Carbonate Device | Request A Quote | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|
Specifications, terms, and pricing are subject to change. Not all products are available in all countries. Please consult your local sales representative for details.