Search Thermo Fisher Scientific
The LC IsoLink II IRMS System offers streamlined workflows for sensitive, precise, online determination of 13C/12C ratios in a variety of applications.
Catalog Number | Type |
---|---|
IQLAAEGAATFAETMAHX | IRMS |
Combine unmatched robustness, ease-of-use, and superior analytical performance to achieve maximum efficiency in your laboratory with the next generation Thermo Scientific™ LC IsoLink™ II IRMS System. All-in-one, integrated stack design features the innovative Thermo Scientific™ Vanquish™ Core HPLC System. The LC IsoLink II IRMS System offers streamlined workflows for sensitive, precise, online determination of 13C/12C ratios in applications such as food integrity, metabolism and physiology studies, drug origin verification, forensics, archaeology, doping control, and biogeochemistry research.
LC IsoLink II IRMS System highlights
High robustness and productivity
The Thermo Scientific™ LC IsoLink™ II Conversion Interface features new, patented technology and user-friendly design to ensure high robustness, easy and quick maintenance and high throughput. A new cartridge-based conversion reactor can be easily exchanged by users, offering cost-efficient system maintenance. The patented backflush function minimizes flow path blockage and maximizes system uptime.
Operational simplicity
The Vanquish Core HPLC system provides high analytical precision and operational simplicity with features such as long-life injection valve and needle alignment, comprehensive instrument diagnostic and the Thermo Scientific™ Viper™ Finger-tight Fittings for nearly zero dead-volume fluidic connections to ensure exceptional instrument reliability.
Smart workflows driven by Qtegra ISDS Software
The LC IsoLink II IRMS System is driven by Qtegra ISDS Software, featuring integration with Thermo Scientific™ Chromeleon™ Chromatography Data System (CDS) capabilities. The layered approach for data visualization, online data evaluation and tailored data reports offers full control and confidence from sample to result.
Specifications, terms, and pricing are subject to change. Not all products are available in all countries. Please consult your local sales representative for details.
Take advantage of this 20% discount trade-in offer to replace an existing Thermo Scientific LC IsoLink system or an instrument from another vendor and benefit from superior analytical performance of our new LC-IRMS platform.
Tracking origins, tracing identity: the Isotope Hunter follows the clues that history leaves in foods or fibers, liquids or stone. Capture your samples’ stories with the Thermo Scientific Isotope Ratio Mass Spectrometry portfolio.
Share catalog number, name or link