Minimize training, automate workflows, simplify your experience, and improve efficiency with the innovative Thermo Scientific™ Qtegra™ Intelligent Scientific Data Solution™ (ISDS) platform software. This shared software approach provides control and data processing for a range of elemental and isotopic analysis technologies including: ICP-OES, ICP-MS, Noble Gas MS, MC-ICP-MS and HR-IRMS. Users can leverage software familiarity across techniques. Designed for simplified workflow compliance, scalability, compliance and data management, Qtegra ISDS Software provides essential tools for consistent, accurate analysis.
Qtegra ISDS Software is updated and upgraded regularly to reflect real-world changes in analytical demands. Recent upgrades include the optional Thermo Scientific™ ChromControl and npQuant plug-ins to make running applications, such as speciation and nanoparticles, even easier.
Type | Software |
Description | Qtegra ISDS Software |
For Use With (Application) | ICP-OES, ICP-MS, Noble Gas MS, and Isotope Ratio Infrared Spectrometry |
Unit Size | Each |
Catalog Number | Specifications | Unit Size | Price (USD) | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
IQLAAEGABSFAOVMBCZ | Each | Request A Quote | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|
One-click "Get Ready" function with automatic performance checks reduces operator time, freeing up valuable resources and increasing accuracy. Three fail-safe steps from sample introduction to report.
Simple, dynamic workflow and intuitive user interface places controls at the operator's disposal for reduced training and expertise requirements.
Just a few quick steps create an analytical LabBook, the start of the intelligent analysis sequence, with full QA/QC protocols and powerful what-you-see-is-what-you-get reports.