ARL SMS-3300 Automation for Single or Twin OES or XRF
ARL SMS-3300 Automation for Single or Twin OES or XRF
ARL SMS-3300 Automation for Single or Twin OES or XRF
ARL SMS-3300 Automation for Single or Twin OES or XRF
ARL SMS-3300 Automation for Single or Twin OES or XRF
Thermo Scientific™
ARL SMS-3300 Automation for Single or Twin OES or XRF
Drive process control laboratory efficiencies in the metals, mining and cement industries with the Thermo Scientific™ ARL™ SMS-3300 Automation for Single or Twin OES or XRF, a complete laboratory workflow automation solution that offers reduced sample analysis cost, increased sample throughput, better reproducibility and higher quality results.
Have Questions?
Catalog NumberFor Use With (Application)
IQLAAHGABDFAAZMBLTMetal, Mining, Cement
Catalog number IQLAAHGABDFAAZMBLT
Price (USD)
-
Request A Quote
For Use With (Application):
Metal, Mining, Cement
Drive process control laboratory efficiencies in the metals, mining and cement industries with the Thermo Scientific™ ARL™ SMS-3300 Automation for Single or Twin OES or XRF, a complete laboratory workflow automation solution. The automation of single or multiple spectrometers and preparation machines with the ARL SMS-3300 system offers reduced sample analysis cost, increased sample throughput, better reproducibility and higher quality results for users requiring more comprehensive and integrated automation solutions in the metals, mining and cement industries.
The ARL SMS-3300 Automation for Single or Twin OES or XRF provides the ultimate power and flexibility for the automatic, unattended round-the-clock operation of the Thermo Scientific™ ARL™ iSpark and ARL iSpark Plus Optical Emission Spectrometers or the Thermo Scientific™ ARL™ 9900 X-ray Fluorescence Spectrometer.
Main Features
  • Designed for easy integration into fully automatic laboratories.
  • The full system can be mounted in the Thermo Scientific ARL QuantoShelter and placed very close to the production process
  • Tasks are performed in parallel; samples are prepared while other samples are introduced or analyzed; the instrument stand cleaning is performed in hidden time by the spectrometer, independently of the ARL SMS-3300 robot
  • Direct access to the milling machine to reduce processing times for ferrous samples
  • More samples processed with the addition of automatic sample preparation solutions with Thermo Scientific™ ARL™ PrepControl™ system and Thermo Scientific™ ARL™ EasyPrep™ software
  • Factory configured and customized
  • Compact chassis and simple installation
  • Integrated electronics
  • Better analysis dependability and quality
  • Very rapid payback
  • Results available faster
  • Qualified lab personnel released from routine duties
  • Manufactured and maintained according to ISO 9001-2000 procedures
Advanced Technology
State-of-the-art Fanuc industrial robot
OXSAS analytical software
Specifications
DescriptionARL SMS-3300 Robotic-based Automation System for Single or Twin OES or XRF
For Use With (Equipment)Thermo Scientific™ ARL™ iSpark Optical Emission Spectrometer or Thermo Scientific™ ARL™ 9900 X-ray Fluorescence Spectrometer
For Use With (Application)Metal, Mining, Cement
Unit SizeEach
Documents & Downloads
Certificates
Safety Data Sheets
Share catalog number, name or link
1x1 image pixel for data collection