Search Thermo Fisher Scientific
Type | X-ray Diffractometer |
Detector Type | Solid State Pixel Detector |
For Use With (Application) | Cement, Coal and Minerals / Manufacturing and Processing / Geosciences / Materials Science |
Unit Size | Each |
Catalog Number | Specifications | Unit Size | Price (USD) | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
IQLAAHGABLFABAMBOC | Each | Request A Quote | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|
Thermo Fisher Scientific continues to innovate with unique product solutions that support your analytical needs and improve process controls.
ARL X'TRA Companion diffractometer is perfectly designed to be a cost-effective solution for routine QC/QA in industrial laboratories. Thanks to its state-of-the art solid-state 2D detector, high resolution data is collected within minutes.
This robust and accurate benchtop X-ray diffractometer comes with one-click Rietveld quantification capabilities and automated result transmission to a LIMS, and it complies with international X-ray safety regulations.
The Thermo Scientific ARL X’TRA Companion is a simple, easy-to-use benchtop XRD systems for routine analysis and more advanced applications.
From exploration to analysis and processing, either online or in the laboratory, we have everything necessary to meet specifications for quality, environment and productivity. Thermo Scientific mining solutions provide reliable, accurate data and essential information for optimal process control.