Thermo Scientific™
SolstiX XRD Software
Catalog number: IQLAAHGABLFAQLMBLQ
Have Questions?
Thermo Scientific™
SolstiX XRD Software
Catalog number: IQLAAHGABLFAQLMBLQ
Have Questions?
Confidently secure your laboratory data and its integrity using the Thermo Scientific™ SolstiX™ XRD Software with Security Suite. This security package is available with our complete ARL™ EQUINOX X-ray diffractometer range and enables 21 CFR Part 11 compatibility.
Catalog Number
IQLAAHGABLFAQLMBLQ
Unit Size
Each
Price (USD)
Full specifications
DescriptionSolstiX XRD Software
Unit SizeEach
Showing 1 of 1
Catalog NumberSpecificationsUnit SizePrice (USD)
IQLAAHGABLFAQLMBLQFull specifications
EachRequest A Quote
DescriptionSolstiX XRD Software
Unit SizeEach
Showing 1 of 1
The SolstiX™ XRD Software with Security Suite features:
  • Full-system access control to authenticate users
  • True digital signatures to record responsibility and provide data integrity protection
  • Traceability with complete audit trails
  • Original data save option with flexible record storage and retrieval
  • Available for all ARL EQUINOX™ models
Software logs change to files associated with the software, even when SolstiX Software itself is not running because system use and file event logging is output in a custom Thermo Fisher log within the Microsoft™ Event Viewer.
Note: Thermo Scientific™ Security Administration Server is responsible for giving system administrator control over software access, policies and digital signature meanings in an intuitive interface. This server can be run stand-alone on your spectrometer workstation or as a server application to administer SolstiX Software on one or all of the systems in your facility.