Search Thermo Fisher Scientific
Type | WDXRF Spectrometer |
Description | ARL OPTIM'X WDXRF Spectrometer; Configurable |
Unit Size | Each |
Catalog Number | Specifications | Unit Size | Description | Price (USD) | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
IQLAAHGABMFAASMACH | Each | ARL OPTIM'X WDXRF Spectrometer; Configurable | Request A Quote | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|
The ARL OPTIM’X WDXRF Spectrometer analyzes elements from carbon to uranium in a wide variety of samples with accuracy, precision and reliability. WDXRF is a standard test method for analytical laboratories (ASTM and ISO norms for example) by virtue of its simplicity, flexibility, affordability and reliability. The cost per analysis is clearly advantageous over many traditional wet chemical and other spectroscopic methods.
Advanced Technology
Main Features
Many sample types can be analyzed including conductive or non-conductive solids, liquids, loose powders, pressed pellets, fusion beads, pastes, granules and coatings.
Recommended for