Search Thermo Fisher Scientific
Description | OXSAS X-Ray Fluorescence Analytical Software |
Unit Size | Each |
Catalog Number | Specifications | Unit Size | Description | Price (USD) | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
IQLAAHGABUFABXMATY | Each | OXSAS X-Ray Fluorescence Analytical Software | Request A Quote | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|
Ease of Use
Integrated Analytical Assistant guides the creation or extension of methods suggesting best analytical parameters for rapid and accurate analyses.
Automatic Method Choice may be used to select the best analyical parameters set without user intervention.
Routine analysis function allows performance of analysis with minimal data-entry, e.g., single sample identity.
One click creation of a batch with all samples requiring analysis.
Simple definition and comprehensive operation of sample batches supports prioritization of samples.
Familiar User Interface
Comprehensive post-treatment of results is available, including export and Microsoft™ Excel support.
Integrated Microsoft SQL Server Express relational database which stores set up and analyses data.
Export and import facilities allow easy transfer of calibration standards, methods or grades from one instrument to another.
Download software updates from OXSAS website offered regularly.
Effective monitoring and maintenance tools maximizes the system uptime.
Real on-line integrated SPC.
SCT Manager: Overview of the analytical capability of the instrument and of each method any time.
Measurement Uncertainty and Conformance Probability on unknown samples analysis.
Fast Calibration
Fast calibration with multiple analyte curve display, instant base curve calculation, templates for edition and measurement of calibration standards.
Calibration curve determination using multi-variable regression with a range of correction models and integrated theoretical alphas calculation.
Available Options
The following optional standard-less analysis packages offer additional analysis versatility:
Thermo Scientific™ QuantAS™ Semi-Quantitative Analysis Package.
Thermo Scientific™ UniQuant™ for determination of up to 79 elements in an unknown sample without the need for specific calibration, effectively eliminating the need for certified materials.
Other Options
ARLcom: Software package for the transmission of results using network and serial communication via Local Area Network to computer applications and printers using TCP/IP or to files.
Remote Sample Definition: Allows samples registration from an external application which can be located anywhere on the network.
Automation: Supports the Thermo Scientific SMS system for full automation of the sample analysis.
Charge correction feature allows the calculation of the weight of materials to be added to a furnace to bring an out-of-specification charge within limits.
SPC-Full: On-line Statistical Process Control, Full Graphical Package Extension.
Compatible with:
Thermo Scientific™ ARL™ PERFORM'X Series, Thermo Scientific™ ARL™ OPTIM'X Series and Thermo Scientific™ ARL™ 9900 IntelliPower Series