Search Thermo Fisher Scientific
Catalog Number | Type |
---|---|
IQLAAMGAATFARHMBLH | MC-ICP-MS |
Perform routine isotope analysis or explore novel isotopes in unknown materials using the Thermo Scientific Neoma multicollector ICP-MS (MC-ICP-MS). This ICP-MS system builds on our multicollector technology to deliver all the advantages of high-precision isotope ratio analysis, with none of the limitations. It includes state-of-the art software and hardware, features flexible and sensitive detector technology, a sensitive ion source and excellent mass resolving power, enabling challenging isotope analyses.
Now Neoma MC-ICP-MS comes with the upgrade pathway to Thermo Scientific™ Neoma™ MS/MS MC-ICP-MS, the first dedicated collision/reaction cell MC-ICP-MS with unique pre-cell mass filtering technology. Allowing you to separate out isobaric interferences and opening up new avenues in the field of laser ablation MC-ICP-MS and beyond.
Technology that transforms your science
Building on the strength of our multicollector technology, the Neoma MC-ICP-MS system blends the cutting-edge and field-proven technologies found in Thermo Scientific high-resolution isotope ratio mass spectrometers and Thermo Scientific™ iCAP™ Qnova ICP-MS systems:
High-precision isotope ratio analysis, simplified
Break through the noise
There are some situations when even the highest resolution is not enough. Isobaric interferences cannot be resolved even with the best ICP-MS instruments available. Thermo Scientific™ Neoma™ MS/MS MC-ICP-MS provides the solution.
For users familiar with triple quad ICP-MS technology, the Neoma MS/MS MC-ICP-MS represents the next leap forward in terms of precision and accuracy. With a similar structure of filter, react, separate, the Neoma MS/MS MC-ICP-MS allows for the effective removal of isobaric interferences without compromising sensitivity, with the additional benefit of simultaneous isotope analysis, for ultimate precision and accuracy of your isotope ratio.
Find out about the technology contained within Neoma MC-ICP-MS and see some examples of MC-ICP-MS for specific applications.
Receive information on products, events, and more to help with your geosciences exploration and analysis.
Share catalog number, name or link