Search Thermo Fisher Scientific
Separate out isobaric interferences to achieve new levels of precision and accuracy in your isotope research. There are some situations when even the highest resolution is not enough and isobaric interferences cannot be resolved, even with the best ICP-MS instruments available. Thermo Scientific™ Neoma™ MS/MS MC-ICP-MS provides the solution.
The Neoma MS/MS MC-ICP-MS is the first dedicated collision/reaction cell MC-ICP-MS with unique pre-cell mass filtering technology, allowing you to separate out isobaric interferences and opening up new avenues in the field of laser ablation MC-ICP-MS and beyond.
Modes:
Applications:
Technology options: