Search Thermo Fisher Scientific
Description | Includes: CD containing a “NanoDrop 2000 series IQ/OQ Procedure and Forms” pdf document, Calibration Check Fluid, Pedestal Reconditioning Kit - Used for reconditioning the sample measurement pedestals to a hydrophobic state, The kit includes a vial of specially formulated compound and a supply of convenient applicators. |
For Use With (Equipment) | NanoDrop 2000/2000C Spectrophotometers |
Includes | CD containing a ''NanoDrop 2000 series IQ/OQ Procedure and Forms'' pdf document, Calibration Check Fluid, Pedestal Reconditioning Kit - Used for reconditioning the sample measurement pedestals to a hydrophobic state, The kit includes a vial of specially formulated compound and a supply of convenient applicators. |
Unit Size | Each |
Catalog Number | Specifications | Unit Size | Price (USD) | Availability | Quantity | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
IQOQ-2000-V131 | Each | 1,630.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|
IQ/OQ Document Kit
Ensures your instrument is installed correctly and working within manufacturer’s specifications.
Includes a USB stick and CD with IQ/OQ procedure, CHEM-PR1-KIT, and CHEM-CF-1.