Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Search Thermo Fisher Scientific
Description | NanoDrop 8000 IQ/OQ Kit |
For Use With (Equipment) | NanoDrop 8000 Spectrophotometers |
Includes | CD containing a ''NanoDrop 8000 IQ/OQ Procedure and Forms'' pdf document, ND 8000 Calibration Check Kit, Pedestal Reconditioning Kit - Used for reconditioning the sample measurement pedestals to a hydrophobic state, The kit includes a vial of specially formulated compound and a supply of convenient applicators. |
Unit Size | Each |
Catalog Number | Specifications | Unit Size | Price (TWD) | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
IQOQ-8000-V210 | Each | Contact Us › | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|
IQ/OQ Document Kit
Ensures your instrument is installed correctly and working within manufacturer’s specifications.
Includes a USB stick and CD with IQ/OQ procedure, CHEM-PR1-KIT, and CHEM-CF-8.