Invitrogen™

iPrep™ Cartridge Rack

Catalog number: IS10102
Have Questions?
Invitrogen™

iPrep™ Cartridge Rack

Catalog number: IS10102
Have Questions?
Catalog Number
IS10102
also known as IS-10102
Unit Size
1 rack (s)
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
IS10102
also known as IS-10102
1 rack (s)
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
The iPrep™ Cartridge Rack is a stainless steel, alluminium alloy rack that holds the pre-filled carteridges during purification. The rack is designed to fit onto the plaform in only one orientation (indicated with an arrow). The racks dimensions are 10.8 inches (l) x 5.5 inches (w) x 2.3 inches (d).
Each iPrep Instrument includes 1 iPRep Carteridge Rack. Additional racks can be ordered for faster instrument set up.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

For Use With (Equipment)
iPrep™
Product Line
iPrep™
Quantity
1 Rack
Shipping Condition
Room Temperature

Contents & Storage

Store at room temperature.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text