Thermo Scientific™
ISQ™ 7610 Single Quadrupole GC-MS
Catalog number: ISQ7610-STNOVPI
Have Questions?
Thermo Scientific™
ISQ™ 7610 Single Quadrupole GC-MS
Catalog number: ISQ7610-STNOVPI
Have Questions?
To stay ahead, analytical testing laboratories need the ultimate confidence of a GC-MS system that easily and reliably produces trusted results, day after day. For this, you can count on the Thermo Scientific™ ISQ™ 7610 Single Quadrupole GC-MS System. Simplified operation, automated workflows, and extended dynamic range deliver consistent results from system to system in every laboratory. Thermo Scientific™ NeverVent™ technology, extended-life detector, and intelligent software eliminate unnecessary downtime to maximize sample throughput. To ensure you are ready for any analytical challenge, the system is upgradeable from entry-level to advanced configurations. Now you can take the lead with rapid return on investment (ROI) for your regulated GC-MS analyses.
ISQ 7610 GC-MS features | Build your GC-MS system | Related products
Catalog Number
ISQ7610-STNOVPI
Unit Size
Each
Description
ISQ 7610 Small Turbo, No VPI with ExtractaBrite
Source Type
Ultra-Inert ExtractaBrite Ion Source
Price (USD)
Full specifications
Mass Range1.2–1100 u with Unit Mass Resolution
TypeSingle Quadrupole GC-MS
DetectorsTriple off-axis Thermo Scientific™ DynaMax™ XTEND XR detection system, with off-axis 10 kV dynode, discrete dynode electron multiplier and electrometer, linear from 0 to 110 μA
ModesElectron Impact Ionization (EI), with Full Scan (FS), SIM, and FS/SIM, Simultaneous within the same injection
Scanning CapabilitiesFast Quadrupole Scanning up to 20,000 U/s
Source TypeUltra-Inert ExtractaBrite Ion Source
Item DescriptionScalable GC-MS platform, fully upgradable from a base configuration to an advanced highly sensitive configuration +
DescriptionISQ 7610 Small Turbo, No VPI with ExtractaBrite
Mass StabilityBetter than 0.1 u/48 hours/∆T ≤2 K
Dimensions46 x 36 x 69 cm (18 x 14 x 27 in) (H x W x D)
Mass AnalyzerDual-stage mass filter with off-axis ion guide pre-filter for noise reduction and solid, homogeneous non- coated, maintenance-free quadrupole rods
Unit SizeEach
Catalog Number
Unit Size
Each
Description
ISQ 7610 Large Turbo VPI with ExtractaBrite
Source Type
Ultra-Inert ExtractaBrite Ion Source
Price (USD)
Full specifications
Mass Range1.2–1100 u with Unit Mass Resolution
TypeSingle Quadrupole GC-MS
DetectorsTriple off-axis Thermo Scientific™ DynaMax™ XTEND XR detection system, with off-axis 10 kV dynode, discrete dynode electron multiplier and electrometer, linear from 0 to 110 μA
ModesElectron Impact Ionization (EI), with Full Scan (FS), SIM, and FS/SIM, Simultaneous within the same injection, Chemical Ionization (CI), with Positive Ion (PCI) and Negative Ion (NCI) Chemical Ionization, Interchangeable without breaking the vacuum
Scanning CapabilitiesFast Quadrupole Scanning up to 20,000 U/s
Source TypeUltra-Inert ExtractaBrite Ion Source
Item DescriptionScalable GC-MS platform, fully upgradable from a base configuration to an advanced highly sensitive configuration
DescriptionISQ 7610 Large Turbo VPI with ExtractaBrite
Mass StabilityBetter than 0.1 u/48 hours/∆T ≤2 K
Dimensions46 x 36 x 69 cm (18 x 14 x 27 in) (H x W x D)
Mass AnalyzerDual-stage mass filter with off-axis ion guide pre-filter for noise reduction and solid, homogeneous non- coated, maintenance-free quadrupole rods
Unit SizeEach
Catalog Number
Unit Size
Each
Description
ISQ 7610 Large Turbo, No VPI with AEI
Source Type
Ultra-Inert ExtractaBrite Ion Source
Price (USD)
Full specifications
Mass Range1.2–1100 u with Unit Mass Resolution
TypeSingle Quadrupole GC-MS
DetectorsTriple off-axis Thermo Scientific™ DynaMax™ XTEND XR detection system, with off-axis 10 kV dynode, discrete dynode electron multiplier and electrometer, linear from 0 to 110 μA
ModesElectron Impact Ionization (EI), with Full Scan (FS), SIM, and FS/SIM, Simultaneous within the same injection, Chemical Ionization (CI), with Positive Ion (PCI) and Negative Ion (NCI) Chemical Ionization, Interchangeable without breaking the vacuum
Scanning CapabilitiesFast Quadrupole Scanning up to 20,000 U/s
Source TypeUltra-Inert ExtractaBrite Ion Source
Item DescriptionScalable GC-MS platform, fully upgradable from a base configuration to an advanced highly sensitive configuration
DescriptionISQ 7610 Large Turbo, No VPI with AEI
Mass StabilityBetter than 0.1 u/48 hours/∆T ≤2 K
Dimensions46 x 36 x 69 cm (18 x 14 x 27 in) (H x W x D)
Mass AnalyzerDual-stage mass filter with off-axis ion guide pre-filter for noise reduction and solid, homogeneous non- coated, maintenance-free quadrupole rods
Unit SizeEach
Catalog Number
Unit Size
Each
Description
ISQ 7610 Large Turbo, No VPI with ExtractaBrite
Source Type
Ultra-Inert ExtractaBrite Ion Source
Price (USD)
Full specifications
Mass Range1.2–1100 u with Unit Mass Resolution
TypeSingle Quadrupole GC-MS
DetectorsTriple off-axis Thermo Scientific™ DynaMax™ XTEND XR detection system, with off-axis 10 kV dynode, discrete dynode electron multiplier and electrometer, linear from 0 to 110 μA
ModesElectron Impact Ionization (EI), with Full Scan (FS), SIM, and FS/SIM, Simultaneous within the same injection
Scanning CapabilitiesFast Quadrupole Scanning up to 20,000 U/s
Source TypeUltra-Inert ExtractaBrite Ion Source
Item DescriptionScalable GC-MS platform, fully upgradable from a base configuration to an advanced highly sensitive configuration
DescriptionISQ 7610 Large Turbo, No VPI with ExtractaBrite
Mass StabilityBetter than 0.1 u/48 hours/∆T ≤2 K
Dimensions46 x 36 x 69 cm (18 x 14 x 27 in) (H x W x D)
Mass AnalyzerDual-stage mass filter with off-axis ion guide pre-filter for noise reduction and solid, homogeneous non- coated, maintenance-free quadrupole rods1.2–1100 u with Unit Mass Resolution
Unit SizeEach
Catalog Number
Unit Size
Each
Description
ISQ 7610 Large Turbo, No VPI with AEI
Source Type
Advanced Electron Ionization (AEI) Source for Ultra-Sensitivity and Robustness
Price (USD)
Full specifications
Mass Range1.2–1100 u with unit mass resolution
TypeSingle Quadrupole GC-MS
DetectorsTriple off-axis Thermo Scientific™ DynaMax™ XTEND XR detection system, with off-axis 10 kV dynode, discrete dynode electron multiplier and electrometer, linear from 0 to 110 μA
ModesElectron Impact Ionization (EI), with Full Scan (FS), SIM, and FS/SIM, Simultaneous within the same injection
Scanning CapabilitiesFast Quadrupole Scanning up to 20,000 U/s
Source TypeAdvanced Electron Ionization (AEI) Source for Ultra-Sensitivity and Robustness
Item DescriptionScalable GC-MS platform, fully upgradable from a base configuration to an advanced highly sensitive configuration
DescriptionISQ 7610 Large Turbo, No VPI with AEI
Mass StabilityBetter than 0.1 u/48 hours/∆T ≤2 K
Dimensions46 x 36 x 69 cm (18 x 14 x 27 in) (H x W x D)
Mass AnalyzerDual-stage mass filter with off-axis ion guide pre-filter for noise reduction and solid, homogeneous non- coated, maintenance-free quadrupole rods
Unit SizeEach
Showing 5 of 5
Catalog NumberSpecificationsUnit SizeDescriptionSource TypePrice (USD)
ISQ7610-STNOVPIFull specifications
EachISQ 7610 Small Turbo, No VPI with ExtractaBriteUltra-Inert ExtractaBrite Ion SourceRequest A Quote
Mass Range1.2–1100 u with Unit Mass Resolution
TypeSingle Quadrupole GC-MS
DetectorsTriple off-axis Thermo Scientific™ DynaMax™ XTEND XR detection system, with off-axis 10 kV dynode, discrete dynode electron multiplier and electrometer, linear from 0 to 110 μA
ModesElectron Impact Ionization (EI), with Full Scan (FS), SIM, and FS/SIM, Simultaneous within the same injection
Scanning CapabilitiesFast Quadrupole Scanning up to 20,000 U/s
Source TypeUltra-Inert ExtractaBrite Ion Source
Item DescriptionScalable GC-MS platform, fully upgradable from a base configuration to an advanced highly sensitive configuration +
DescriptionISQ 7610 Small Turbo, No VPI with ExtractaBrite
Mass StabilityBetter than 0.1 u/48 hours/∆T ≤2 K
Dimensions46 x 36 x 69 cm (18 x 14 x 27 in) (H x W x D)
Mass AnalyzerDual-stage mass filter with off-axis ion guide pre-filter for noise reduction and solid, homogeneous non- coated, maintenance-free quadrupole rods
Unit SizeEach
ISQ7610-VPIFull specifications
EachISQ 7610 Large Turbo VPI with ExtractaBriteUltra-Inert ExtractaBrite Ion SourceRequest A Quote
Mass Range1.2–1100 u with Unit Mass Resolution
TypeSingle Quadrupole GC-MS
DetectorsTriple off-axis Thermo Scientific™ DynaMax™ XTEND XR detection system, with off-axis 10 kV dynode, discrete dynode electron multiplier and electrometer, linear from 0 to 110 μA
ModesElectron Impact Ionization (EI), with Full Scan (FS), SIM, and FS/SIM, Simultaneous within the same injection, Chemical Ionization (CI), with Positive Ion (PCI) and Negative Ion (NCI) Chemical Ionization, Interchangeable without breaking the vacuum
Scanning CapabilitiesFast Quadrupole Scanning up to 20,000 U/s
Source TypeUltra-Inert ExtractaBrite Ion Source
Item DescriptionScalable GC-MS platform, fully upgradable from a base configuration to an advanced highly sensitive configuration
DescriptionISQ 7610 Large Turbo VPI with ExtractaBrite
Mass StabilityBetter than 0.1 u/48 hours/∆T ≤2 K
Dimensions46 x 36 x 69 cm (18 x 14 x 27 in) (H x W x D)
Mass AnalyzerDual-stage mass filter with off-axis ion guide pre-filter for noise reduction and solid, homogeneous non- coated, maintenance-free quadrupole rods
Unit SizeEach
ISQ7610-VPICIFull specifications
EachISQ 7610 Large Turbo, No VPI with AEIUltra-Inert ExtractaBrite Ion SourceRequest A Quote
Mass Range1.2–1100 u with Unit Mass Resolution
TypeSingle Quadrupole GC-MS
DetectorsTriple off-axis Thermo Scientific™ DynaMax™ XTEND XR detection system, with off-axis 10 kV dynode, discrete dynode electron multiplier and electrometer, linear from 0 to 110 μA
ModesElectron Impact Ionization (EI), with Full Scan (FS), SIM, and FS/SIM, Simultaneous within the same injection, Chemical Ionization (CI), with Positive Ion (PCI) and Negative Ion (NCI) Chemical Ionization, Interchangeable without breaking the vacuum
Scanning CapabilitiesFast Quadrupole Scanning up to 20,000 U/s
Source TypeUltra-Inert ExtractaBrite Ion Source
Item DescriptionScalable GC-MS platform, fully upgradable from a base configuration to an advanced highly sensitive configuration
DescriptionISQ 7610 Large Turbo, No VPI with AEI
Mass StabilityBetter than 0.1 u/48 hours/∆T ≤2 K
Dimensions46 x 36 x 69 cm (18 x 14 x 27 in) (H x W x D)
Mass AnalyzerDual-stage mass filter with off-axis ion guide pre-filter for noise reduction and solid, homogeneous non- coated, maintenance-free quadrupole rods
Unit SizeEach
ISQ7610-NOVPIFull specifications
EachISQ 7610 Large Turbo, No VPI with ExtractaBriteUltra-Inert ExtractaBrite Ion SourceRequest A Quote
Mass Range1.2–1100 u with Unit Mass Resolution
TypeSingle Quadrupole GC-MS
DetectorsTriple off-axis Thermo Scientific™ DynaMax™ XTEND XR detection system, with off-axis 10 kV dynode, discrete dynode electron multiplier and electrometer, linear from 0 to 110 μA
ModesElectron Impact Ionization (EI), with Full Scan (FS), SIM, and FS/SIM, Simultaneous within the same injection
Scanning CapabilitiesFast Quadrupole Scanning up to 20,000 U/s
Source TypeUltra-Inert ExtractaBrite Ion Source
Item DescriptionScalable GC-MS platform, fully upgradable from a base configuration to an advanced highly sensitive configuration
DescriptionISQ 7610 Large Turbo, No VPI with ExtractaBrite
Mass StabilityBetter than 0.1 u/48 hours/∆T ≤2 K
Dimensions46 x 36 x 69 cm (18 x 14 x 27 in) (H x W x D)
Mass AnalyzerDual-stage mass filter with off-axis ion guide pre-filter for noise reduction and solid, homogeneous non- coated, maintenance-free quadrupole rods1.2–1100 u with Unit Mass Resolution
Unit SizeEach
ISQ7610-AEIFull specifications
EachISQ 7610 Large Turbo, No VPI with AEIAdvanced Electron Ionization (AEI) Source for Ultra-Sensitivity and RobustnessRequest A Quote
Mass Range1.2–1100 u with unit mass resolution
TypeSingle Quadrupole GC-MS
DetectorsTriple off-axis Thermo Scientific™ DynaMax™ XTEND XR detection system, with off-axis 10 kV dynode, discrete dynode electron multiplier and electrometer, linear from 0 to 110 μA
ModesElectron Impact Ionization (EI), with Full Scan (FS), SIM, and FS/SIM, Simultaneous within the same injection
Scanning CapabilitiesFast Quadrupole Scanning up to 20,000 U/s
Source TypeAdvanced Electron Ionization (AEI) Source for Ultra-Sensitivity and Robustness
Item DescriptionScalable GC-MS platform, fully upgradable from a base configuration to an advanced highly sensitive configuration
DescriptionISQ 7610 Large Turbo, No VPI with AEI
Mass StabilityBetter than 0.1 u/48 hours/∆T ≤2 K
Dimensions46 x 36 x 69 cm (18 x 14 x 27 in) (H x W x D)
Mass AnalyzerDual-stage mass filter with off-axis ion guide pre-filter for noise reduction and solid, homogeneous non- coated, maintenance-free quadrupole rods
Unit SizeEach
Showing 5 of 5
Increase instrument uptime
Eliminate unnecessary, unplanned instrument downtime and maximize productivity to achieve unprecedented efficiency, day after day. The ISQ 7610 Single Quadrupole GC-MS System combines robustness with the ability to change the GC column and clean the ion source without interrupting your analytical workflows.
Monitor key aspects of your GC-MS system
With Thermo Scientific™ SmartStatus™ instrument tracking within Chromeleon CDS, quickly see whether instruments are ready to run samples, if maintenance is required or if consumables should be replaced. The fully customizable software enables you to make data-driven decisions on when to perform maintenance and avoid unnecessary downtime.
Maximize sample throughput
When high sample throughput is essential, the system delivers results on time and with ease. Automated workflows and simplified instrument operation ensure every user produces consistent results, sample after sample. Extended linear range enables method consolidation so you can analyze more compounds at varying concentrations in a single run.
Realize rapid return on investment
Ensuring your system delivers results as soon as it’s installed is necessary to achieve rapid ROI. With built-in intelligence that simplifies instrument setup, analytical methods, and everyday operation, the ISQ 7610 Single Quadrupole GC-MS System is designed for accelerated deployment. Reduced needs for operator training and faster time to full productivity together with maximum sample throughput provide fast return on your instrument investment.