Anza™ 87 Eco47I
Anza™ 87 Eco47I
Invitrogen™
Anza™ 87 Eco47I
5' G ↓ G W C C 3'3' C C W G ↑ G 5'Invitrogen™ Anza™ 87 Eco47I is aRead more
Have Questions?
Catalog number IVGN0876
Price (USD)/ 1500 units
108.00
-
Add to cart
Price (USD)/ 1500 units
108.00
Add to cart
Anza™ 87 Eco47I
Catalog numberIVGN0876
Price (USD)/ 1500 units
108.00
-
Add to cart
5'    G ↓ G    W    C    C    3'
3'    C    C    W    G ↑ G    5'
Invitrogen™ Anza™ 87 Eco47I is a restriction enzyme that cuts DNA at this recognition site: G^GWCC, completely digesting the DNA in 15 minutes at 37°C. Isoschizomers include: AvaII, Bme18I, SinI, VpaK11BI.
For superior convenience, a single buffer and protocol are used with all Anza™ restriction enzymes. The single Anza buffer allows multiple restriction enzymes to be used simultaneously in one reaction. This helps eliminate the need for sequential digests and the DNA clean-up necessary when changing buffers. Additionally, the Anza buffer and restriction enzymes have been designed for flexibility in digestion times. Digestions of up to 16 hours can be carried out without star activity.
Convenient—all Anza enzymes are 100% functional in the single Anza buffer
Fast—complete digestion in 15 minutes
Flexible—overnight digestion without star activity
Simple—a single digestion protocol for all DNA types
Complete system—incorporates Anza DNA Modifying Enzymes for a complete cloning experience
Anza DNA Modifying Enzymes include the Anza T4 DNA Ligase Master Mix, Anza Alkaline Phosphatase Kit, Anza T4 PNK Kit, Anza DNA Blunt End Kit, and Anza DNA End Repair Kit. Use these enzymes for a complete cloning workflow.
Anza Red Buffer, which contains a density gradient and tracking dyes, is supplied for added convenience. Digestion reactions are as efficient in the Anza Red Buffer as in the Anza Buffer. Use of the Anza Red Buffer helps reduce pipetting steps when preparing multiple digestion reactions and a subsequent step includes agarose gel electrophoresis. For applications that require product analysis by fluorescence excitation, the colorless Anza Buffer without the tracking dyes is recommended.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Shipping ConditionApproved for shipment on Wet or Dry Ice
Methylation SensitivityCpG Methylation-Sensitive, Not CpG Methylation-Sensitive, Not Dam Methylation-Sensitive
EnzymeEco47I
Compatible BufferAnza™ 10X Red Buffer, Anza™ 10X Buffer
Sensitive to Heat InactivationYes
Optimal Reaction Temperature37°C
Type IIS RENo
Quantity1500 units
Product TypeRestriction Enzyme Cloning System
Research CategoryTraditional Cloning
Product LineAnza™
Unit Size1500 units
Contents & Storage
75 μL Anza 87 Eco47I
500 μL Anza Buffer (10X)
500 μL Anza Red Buffer (10X)
Store at -20°C.