Invitrogen™

Anza™ 5-Pack Starter Kit

Catalog number: IVGN3004
Have Questions?
Invitrogen™

Anza™ 5-Pack Starter Kit

Catalog number: IVGN3004
Have Questions?
Catalog Number
IVGN3004
Unit Size
1 kit
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
IVGN30041 kit
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
The Invitrogen™ Anza™ 5-Pack Starter Kit includes the 5 common restriction enzymes, 2 DNA modifying enzymes, and buffers listed below:

• Anza™ 11 EcoRI
• Anza™ 5 BamHI
• Anza™ 1 NotI
• Anza™ 16 HindIII
• Anza™ 12 XbaI
• Anza™ T4 DNA Ligase Master Mix
• Anza™ Alkaline Phosphatase
• Anza™ Buffer (10X)
• Anza™ Red Buffer (10X)

For a kit with 10 enzymes, see the Anza™ 10-Pack Starter Kit.

The Anza™ Restriction Enzyme Cloning System employs a single buffer and protocol for all Anza restriction enzymes. The single Anza buffer allows multiple restriction enzymes to be used simultaneously in one reaction. This eliminates the need for sequential digests and the DNA clean-up necessary when changing buffers. Additionally, the Anza buffer and restriction enzymes have been designed for flexibility in digestion times. Digestions of up to 16 hours can be carried out without star activity.

Convenient—all Anza enzymes are 100% functional in the single Anza buffer
Fast—complete digestion in 15 minutes
Flexible—overnight digestion without star activity
Simple—a single digestion protocol for all DNA types
Complete system—incorporates Anza DNA Modifying Enzymes for a complete cloning experience

Anza Red Buffer, which contains a density gradient and tracking dyes, is supplied for added convenience. Digestion reactions are as efficient in the Anza Red Buffer as in the Anza Buffer. Use of the Anza Red Buffer helps reduce pipetting steps when preparing multiple digestion reactions and a subsequent step includes agarose gel electrophoresis. For applications that require product analysis by fluorescence excitation, the colorless Anza Buffer without the tracking dyes is recommended.

The Anza 5-pk Starter Kit is a subset of the Anza Restriction Enzyme Cloning System, unifying the traditional cloning processes. For more information on the system, visit www.thermofisher.com/anza.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Shipping Condition
Approved for shipment on Wet or Dry Ice
Methylation Sensitivity
Methylation Sensitivities Varies by Enzyme
Enzyme
HindIII, NotI, Alkaline Phosphatase, XbaI, T4 DNA Ligase, BamHI, EcoRI
Compatible Buffer
Anza™ 10X Red Buffer, Anza™ 10X Buffer
Type IIS RE
No
Product Type
Starter Kit
Research Category
Traditional Cloning
Product Line
Anza™

Contents & Storage

• 20 μL each of Anza 11 EcoRI, Anza 5 BamHI, Anza 1 NotI, Anza 16 HindIII, Anza 12 XbaI
• 100 μL Anza T4 DNA Ligase Master Mix
• 20 μL Anza Alkaline Phosphatase
• 500 μL Anza Buffer (10X)
• 500 μL Anza Red Buffer (10X)

Store at -5 to -30°C.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text