Thermo Scientific Chemicals

Bacteriological Agar, Ultrapure, Thermo Scientific Chemicals

Catalog number: J10906.P5
5 lb., Each
Thermo Scientific Chemicals

Bacteriological Agar, Ultrapure, Thermo Scientific Chemicals

Catalog number: J10906.P5
5 lb., Each
Quantity
Catalog number: J10906.P5
Price (USD)
Have Questions?

Chemical Identifiers

CAS
9002-18-0
IUPAC Name
2-({4-hydroxy-3-methyl-2,6-dioxabicyclo[3.2.1]octan-8-yl}oxy)-6-(hydroxymethyl)-4-methoxyoxane-3,5-diol
Molecular Formula
C14H24O9
InChI Key
GYYDPBCUIJTIBM-UHFFFAOYNA-N
SMILES
COC1C(O)C(CO)OC(OC2C3COC2C(O)C(C)O3)C1O
Form
White to light cream-colored powder
Assay (unspecified)
Gel pH (1.5%) after autoclaving: 6.0 - 7.5
Assay (unspecified)
Gel Point (1.5%): 32 - 38?C
Assay (unspecified)
Gel Strength (1.5%): 550 - 950 gm/cm²
Melting Point
(1.5%): 80 - 90?C

Description

Used extensively as a gelling agent in the preparation of bacteriological culture media. Its main attributes are superior transparency, very reliable reproducibility and absence of inhibitors capable of hindering optimal development of microorganisms. Extraneous matter reduced to a minimum.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Superior transparency
  • Reliable reproducibility
  • Absence of inhibitors capable of hindering optimal development of microorganisms
  • Extraneous matter reduced to minimum
  • Format: White to light cream-colored powder
  • CAS: 9002-18-0
  • MDL Number: MFCD00081288
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text