Noble Agar, Ultrapure, Thermo Scientific Chemicals
Noble Agar, Ultrapure, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Noble Agar, Ultrapure, Thermo Scientific Chemicals

Gelling agent essentially free of impurities. Thermo Scientific™ Noble Agar (Ultrapure) is mainly used in biochemistry and molecular biology procedures where increased purity is required.
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
1 kg
Catalog number J10907.36
Price (USD)/ Each
208.65
Online exclusive
232.00 
Save 23.35 (10%)
-
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (USD)/ Each
208.65
Online exclusive
232.00 
Save 23.35 (10%)
Add to cart
Noble Agar, Ultrapure, Thermo Scientific Chemicals
Catalog numberJ10907.36
Price (USD)/ Each
208.65
Online exclusive
232.00 
Save 23.35 (10%)
-
Add to cart
Chemical Identifiers
CAS9002-18-0
IUPAC Name2-({4-hydroxy-3-methyl-2,6-dioxabicyclo[3.2.1]octan-8-yl}oxy)-6-(hydroxymethyl)-4-methoxyoxane-3,5-diol
Molecular FormulaC14H24O9
InChI KeyGYYDPBCUIJTIBM-UHFFFAOYNA-N
SMILESCOC1C(O)C(CO)OC(OC2C3COC2C(O)C(C)O3)C1O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)Gel pH (1.5%): 5.0 - 7.0
Assay (unspecified)Gel Point (1.5%): 32? - 37.5?C
Assay (unspecified)Gel Strength (1.5%): ≥700 gm/cm²
Ash≤ 1.6%
CommentEEO (-Mr): ≤ 0.450 (pH 8.4)
View more

Gelling agent essentially free of impurities. Used mainly in biochemistry and molecular biology procedures wherever increased purity is required.

Typical Agar Preparation:

  • Plates: 15 g/L of media; 30-35 ml per 90 mm plate
  • Top Agar: 7 g/L of media

Autoclave for 20 minutes and swirl gently to evenly distribute the agar. If antibiotics are to be added, allow the agar to cool to 50°C. When pouring plates, gently swirl the medium. A Bunsen burner flame can be passed over the surface of the plates to remove any air bubbles. After the medium has hardened, invert the plates and store at 4°C.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Format: Pale cream powder
  • CAS: 9002-18-0
  • MDL Number: MFCD00081288
Typical Agar Preparation
Plates: 15g/L of media; 30 to 35mL per 90mm plate
Top Agar: 7g/L of media
Autoclave for 20 minutes and swirl gently to evenly distribute the agar. If antibiotics are to be added, allow the agar to cool to 50°C. When pouring plates, gently swirl the medium. A Bunsen burner flame can be passed over the surface of the plates to remove any air bubbles. After the medium has hardened, invert the plates and store at 4°C
RUO – Research Use Only