Thermo Scientific Chemicals

Ammonium Sulfate, ACS, ≥99%, Ultrapure, Thermo Scientific Chemicals

Catalog number: J11254.A1
1 kg, Plastic bottle, Each
Thermo Scientific Chemicals

Ammonium Sulfate, ACS, ≥99%, Ultrapure, Thermo Scientific Chemicals

Catalog number: J11254.A1
1 kg, Plastic bottle, Each
Quantity
Packaging
Catalog number: J11254.A1
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7783-20-2
IUPAC Name
sulfuric acid diamine
Molecular Formula
H8N2O4S
InChI Key
BFNBIHQBYMNNAN-UHFFFAOYSA-N
SMILES
N.N.OS(O)(=O)=O
Form
White crystalline powder
Assay (unspecified)
≥ 99.0%
pH
(5%, H₂O, 25?C): 5.0 - 6.0
Trace Metal
Heavy Metals (Pb): ≤ 5 ppm
Trace Metal
Iron (Fe): ≤ 5 ppm

Description

Used in the purification of enzymes and other proteins by precipitation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text