Aprotinin From Bovine Lung, Thermo Scientific Chemicals
Aprotinin From Bovine Lung, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Aprotinin From Bovine Lung, Thermo Scientific Chemicals

Useful protease inhibitor against kallikrein, trypsin, plasmin and chymotrypsin. Thermo Scientific™ Aprotinin From Bovine Lung is not effective against papain.
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mg
10 mg
100 mg
Catalog number J11388.MB
Price (USD)/ Each
168.00
-
Add to cart
Quantity:
25 mg
Request bulk or custom format
Price (USD)/ Each
168.00
Add to cart
Aprotinin From Bovine Lung, Thermo Scientific Chemicals
Catalog numberJ11388.MB
Price (USD)/ Each
168.00
-
Add to cart
SpecificationsSpecification SheetSpecification Sheet
Concentration1 to 2 mg/mL (Soluble in 0.01 M HEPES)
For Use With (Application)Protease Inhibition
Molecular Weight (g/mol)6511.44
NameAprotinin
pH Range8
View more
A useful protease inhibitor against kallikrein, trypsin, plasmin and chymotrypsin. Not effective against papain.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Activity: 5100 to 7300 Kallikrein inhibitor U/mg protein
  • Molecular Formula: C284H432N84O79S7
  • MDLNumber: MFCD00130541
  • CAS: 9087-70-1
RUO – Research Use Only