Thermo Scientific Chemicals

Aprotinin From Bovine Lung, Thermo Scientific Chemicals

Catalog number: J11388.MC
100 mg, Each
Thermo Scientific Chemicals

Aprotinin From Bovine Lung, Thermo Scientific Chemicals

Catalog number: J11388.MC
100 mg, Each
Quantity
Catalog number: J11388.MC
Price (USD)
Quantity
-
Have Questions?
Assay (unspecified)
Purity (gel filtration): ≥ 95%
Concentration
Working Concentration: Soluble in 0.01M HEPES (pH 8.0)
Form
Lyophilized powder
Assay (unspecified)
Activity: 5,100 - 7,300 Kallikrein Inhibitor units/mg protein
Comment
Avoid repeated freezing and thawing of stock solutions.

Description

A useful protease inhibitor against kallikrein, trypsin, plasmin and chymotrypsin. Not effective against papain.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Activity: 5100 to 7300 Kallikrein inhibitor U/mg protein
  • Molecular Formula: C284H432N84O79S7
  • MDLNumber: MFCD00130541
  • CAS: 9087-70-1
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text