2'-Deoxyguanosine Monohydrate, Synthetic, >98% (Dry Basis), Thermo Scientific Chemicals
2'-Deoxyguanosine Monohydrate, Synthetic, >98% (Dry Basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

2'-Deoxyguanosine Monohydrate, Synthetic, >98% (Dry Basis), Thermo Scientific Chemicals

 
Have Questions?
Quantity:
10 g
Packaging:
Plastic bottle
Catalog number J14304.09
Price (USD)/ Each
827.00
-
Add to cart
Quantity:
10 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)/ Each
827.00
Add to cart
2'-Deoxyguanosine Monohydrate, Synthetic, >98% (Dry Basis), Thermo Scientific Chemicals
Catalog numberJ14304.09
Price (USD)/ Each
827.00
-
Add to cart
Chemical Identifiers
CAS312693-72-4
IUPAC Name2-amino-9-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-6,9-dihydro-1H-purin-6-one hydrate
Molecular FormulaC10H15N5O5
InChI KeyLZSCQUCOIRGCEJ-VDPVQVSMNA-N
SMILESO.NC1=NC2=C(N=CN2[C@H]2C[C@H](O)[C@@H](CO)O2)C(=O)N1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)≥ 98% (dry basis)
Assay (special)Extinction Coefficient: 13.7 X 10³ ? 4% (pH 7.0)
lambda-max (UV-Vis)253 ± 2 nm (pH 7.0)
FormWhite crystalline powder
Identification (FTIR)Nujol mull matches reference scan
View more
 
RUO – Research Use Only