Dithiothreitol, >99.5%, Molecular Biology Grade, Ultrapure, Thermo Scientific Chemicals
Dithiothreitol, >99.5%, Molecular Biology Grade, Ultrapure, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Dithiothreitol, >99.5%, Molecular Biology Grade, Ultrapure, Thermo Scientific Chemicals

Used as protective agent for SH groups
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
100 g
Packaging:
Plastic bottle
Catalog number J15397.03
Price (USD)/ Each
63.90
-
Add to cart
Quantity:
1 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)/ Each
63.90
Add to cart
Dithiothreitol, >99.5%, Molecular Biology Grade, Ultrapure, Thermo Scientific Chemicals
Catalog numberJ15397.03
Price (USD)/ Each
63.90
-
Add to cart
Chemical Identifiers
CAS3483-12-3
IUPAC Name(2R,3R)-1,4-disulfanylbutane-2,3-diol
Molecular FormulaC4H10O2S2
InChI KeyVHJLVAABSRFDPM-IMJSIDKUSA-N
SMILESO[C@@H](CS)[C@@H](O)CS
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)Absorbance (280 nm, 0.1M, 1 cm): ≤ 0.08
Impurity contentOxidized DTT: ≤ 0.3%
Protease activityPasses test
Assay (unspecified)(SH): ≥ 99.5%
DNasePasses test
View more
Used as a protective agent for SH groups.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only