Ethylenediaminetetraacetic acid, (EDTA), 0.5M Solution, Molecular Biology Grade, Ultrapure, Thermo Scientific Chemicals
Ethylenediaminetetraacetic acid, (EDTA), 0.5M Solution, Molecular Biology Grade, Ultrapure, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ethylenediaminetetraacetic acid, (EDTA), 0.5M Solution, Molecular Biology Grade, Ultrapure, Thermo Scientific Chemicals

Chelating agent used in many enzyme buffers and at higher concentrations, as enzyme inactivator
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
500 mL
Packaging:
Nalgene bottle
Catalog number J15694.AE
Price (USD)/ Each
40.65
Online exclusive
45.50 
Save 4.85 (11%)
-
Add to cart
Quantity:
100 mL
Packaging:
Nalgene bottle
Request bulk or custom format
Price (USD)/ Each
40.65
Online exclusive
45.50 
Save 4.85 (11%)
Add to cart
Ethylenediaminetetraacetic acid, (EDTA), 0.5M Solution, Molecular Biology Grade, Ultrapure, Thermo Scientific Chemicals
Catalog numberJ15694.AE
Price (USD)/ Each
40.65
Online exclusive
45.50 
Save 4.85 (11%)
-
Add to cart
Chemical Identifiers
CAS6381-92-6
IUPAC Namedisodium 2-({2-[(carboxylatomethyl)(carboxymethyl)amino]ethyl}(carboxymethyl)amino)acetate dihydrate
Molecular FormulaC10H18N2Na2O10
InChI KeyOVBJJZOQPCKUOR-UHFFFAOYSA-L
SMILESO.O.[Na+].[Na+].OC(=O)CN(CCN(CC(O)=O)CC([O-])=O)CC([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Protease activityPasses test
DNase(exo): Passes test
RNasePasses test
DNase(endo): Passes test
pH8.0 ? 0.5
Chelating agent used in many enzyme buffers and, at higher concentrations, as an enzyme inactivator.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only